Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • New Products
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • TRAPPC10 Antibodies

          Abnova

          TRAPPC10 Polyclonal Antibody, MaxPab™

          View all (8) TRAPPC10 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers

          Cite TRAPPC10 Polyclonal Antibody, MaxPab™

          {{$ctrl.translations['antibody.pdp.skuNotes.additionalInformation']}}:

          {{$ctrl.translations[$ctrl.isSkuNotesCollapsed ? 'antibody.pdp.skuNotes.viewMore' : 'antibody.pdp.skuNotes.viewLess']}}
          • Antibody Testing Data (1)
          TRAPPC10 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          TRAPPC10 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.

          FIGURE: 1 / 1

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          TRAPPC10 Antibody (H00007109-B01P) in WB

          Western Blot analysis of TRAPPC10 expression in transfected 293T cell line by TRAPPC10 MaxPab polyclonal antibody. Lane 1: TMEM1 transfected lysate(138.49 KDa). Lane 2: Non-transfected lysate. . {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          TRAPPC10 Antibody in Western Blot (WB)

          Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.

          TRAPPC10 Polyclonal Antibody, MaxPab™

          Product Details

          H00007109-B01P

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:500-1:1,000
          -
          Product Specifications

          Species Reactivity

          Human

          Host/Isotype

          Mouse / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          TRAPPC10 (NP_003265.3, 1 a.a. approximately 1259 a.a) full-length human protein.
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          1 mg/mL

          Amount

          50 µg

          Purification

          Protein A

          Storage buffer

          PBS, pH 7.4

          Contains

          no preservative

          Storage conditions

          -20°C, Avoid Freeze/Thaw Cycles

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          Product Specific Information

          Product may be used with Western Blot (Transfected lysate).

          The Isotype of this product is composed of an IgG Mixture.

          Immunogen sequence: MDASEEPLPP VIYTMENKPI VTCAGDQNLF TSVYPTLSQQ LPREPMEWRR SYGRAPKMIH LESNFVQFKE ELLPKEGNKA LLTFPFLHIY WTECCDTEVY KATVKDDLTK WQNVLKAHSS VDWLIVIVEN DAKKKNKTNI LPRTSIVDKI RNDFCNKQSD RCVVLSDPLK DSSRTQESWN AFLTKLRTLL LMSFTKNLGK FEDDMRTLRE KRTEPGWSFC EYFMVQEELA FVFEMLQQFE DALVQYDELD ALFSQYVVNF GAGDGANWLT FFCQPVKSWN GLILRKPIDM EKRESIQRRE ATLLDLRSYL FSRQCTLLLF LQRPWEVAQR ALELLHNCVQ ELKLLEVSVP PGALDCWVFL SCLEVLQRIE GCCDRAQIDS NIAHTVGLWS YATEKLKSLG YLCGLVSEKG PNSEDLNRTV DLLAGLGAER PETANTAQSP YKKLKEALSS VEAFEKHYLD LSHATIEMYT SIGRIRSAKF VGKDLAEFYM RKKAPQKAEI YLQGALKNYL AEGWALPITH TRKQLAECQK HLGQIENYLQ TSSLLASDHH LTEEERKHFC QEILDFASQP SDSPGHKIVL PMHSFAQLRD LHFDPSNAVV HVGGVLCVEI TMYSQMPVPV HVEQIVVNVH FSIEKNSYRK TAEWLTKHKT SNGIINFPPE TAPFPVSQNS LPALELYEMF ERSPSDNSLN TTGIICRNVH MLLRRQESSS SLEMPSGVAL EEGAHVLRCS HVTLEPGANQ ITFRTQAKEP GTYTLRQLCA SVGSVWFVLP HIYPIVQYDV YSQEPQLHVE PLADSLLAGI PQRVKFTVTT GHYTIKNGDS LQLSNAEAML ILCQAESRAV VYSNTREQSS EAALRIQSSD KVTSISLPVA PAYHVIEFEL EVLSLPSAPA LGGESDMLGM AEPHRKHKDK QRTGRCMVTT DHKVSIDCPW SIYSTVIALT FSVPFRTTHS LLSSGTRKYV QVCVQNLSEL DFQLSDSYLV DTGDSTDLQL VPLNTQSQQP IYSKQSVFFV WELKWTEEPP PSLHCRFSVG FSPASEEQLS ISLKPYTYEF KVENFFTLYN VKAEIFPPSG MEYCRTGSLC SLEVLITRLS DLLEVDKDEA LTESDEHFST KLMYEVVDNS SNWAVCGKSC GVISMPVAAR ATHRVHMEVM PLFAGYLPLP DVRLFKYLPH HSAHSSQLDA DSWIENDSLS VDKHGDDQPD SSSLKSRGSV HSACSSEHKG LPMPRLQALP AGQVFNSSSG TQVLVIPSQD DHVLEVSVT

          Target Information

          TRAPPC10, also known as TMEM1 (transmembrane protein 1), EHOC1 (epilepsy holoprosencephaly candidate 1 protein) or GT334, is a widely expressed 1,259 amino acid protein that may function in vesicular transport. Despite its name, TRAPPC10 does not contain transmembrane domains. It is the human ortholog of the yeast Trs130 protein and its structure and function appears to be conserved. Localizing to the cis-Golgi apparatus, TRAPPC10 is believed to be involved in transport from the endoplasmic reticulum (ER) to the Golgi functioning as a component of the multisubunit transport protein particle (TRAPP) complex. Mutations in the gene encoding TRAPPC10 may be involved in autoimmune polyglandular disease type 1 or Unverricht-Lundborg disease, an autosomal recessive type of progressive myoclonic epilepsy.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: epilepsy holoprosencephaly candidate-1 protein; putative transmembrane protein; trafficking protein particle complex 10; trafficking protein particle complex subunit 130; trafficking protein particle complex subunit TMEM1; transmembrane protein 1; transport protein particle subunit TMEM1; TRAPP 130 kDa subunit; TRAPP subunit TMEM1; unnamed protein product

          View more View less

          Gene Aliases: EHOC-1; EHOC1; GT334; NEDMISS; TMEM1; TRS130; TRS30

          View more View less

          UniProt ID: (Human) P48553

          View more View less

          Entrez Gene ID: (Human) 7109

          View more View less

          Function(s)
          guanyl-nucleotide exchange factor activity protein binding
          Process(es)
          ER to Golgi vesicle-mediated transport intra-Golgi vesicle-mediated transport vesicle coating early endosome to Golgi transport Golgi vesicle transport
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-b6cf8ccb7-5t5fd:80/100.66.72.150:80.
          git-commit: 76b2fce788d0185f5e22c122d85c407206860d42
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.60.0-2026-offline-prod