Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          • Primary Antibodies ›
          • TNFAIP6 Antibodies

          Proteintech

          TSG-6 Polyclonal Antibody

          5 References
          View all (8) TNFAIP6 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support

          Cite TSG-6 Polyclonal Antibody

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (2)
          TSG-6 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          TSG-6 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.

          FIGURE: 1 / 2

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          TSG-6 Antibody (13321-1-AP) in WB

          NIH/3T3 cells were subjected to SDS PAGE followed by western blot with 13321-1-AP ( TSG-6 antibody at dilution of 1:600 incubated at room temperature for 1.5 hours. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          TSG-6 Antibody in Western Blot (WB)
          TSG-6 Antibody in Western Blot (WB)
          TSG-6 Polyclonal Antibody

          Product Details

          13321-1-AP

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:500-1:1,000
          View 4 publications 4 publications

          Immunohistochemistry (IHC)

          -
          View 1 publication 1 publication
          Product Specifications

          Species Reactivity

          Human, Mouse, Rat

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          TSG-6 Fusion Protein Ag3885 (1-277 aa encoded by BC030205)
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          0.23 mg/mL

          Amount

          34.5 µg

          Purification

          Antigen affinity chromatography

          Storage buffer

          PBS, pH 7.3, with 50% glycerol

          Contains

          0.02% sodium azide

          Storage conditions

          -20°C

          Shipping conditions

          Wet ice (domestic); Dry ice (international)

          Product Specific Information

          Immunogen sequence: MIILIYLFL LLWEDTQGWG FKDGIFHNSI WLERAAGVYH REARSGKYKL TYAEAKAVCE FEGGHLATYK QLEAARKIGF HVCAAGWMAK GRVGYPIVKP GPNCGFGKTG IIDYGIRLNR SERWDAYCYN PHAKECGGVF TDPKQIFKSP GFPNEYEDNQ ICYWHIRLKY GQRIHLSFLD FDLEDDPGCL ADYVEIYDSY DDVHGFVGRY CGDELPDDII STGNVMTLKF LSDASVTAGG FQIKYVAMDP VSKSSQGKNT STTSTGNKNF LAGRFSHL (1-277 aa encoded by BC030205)

          Target Information

          The TSG6 gene is transcribed in normal fibroblasts and activated by binding of the cytokines TNF alpha and IL-1 at AP-1 and NF-IL6 sites in its promoter. TSG-6 is a glycoprotein and a member of the hyaluronan-binding protein family, which includes cartilage link protein, proteoglycan core protein and the adhesion receptor CD44. TSG-6 is highly homologous to CD44, particularly in the hyaluronic acid-binding domain. TSG-6 is found in TNF-treated cells; its expression is rapidly activated by TNF alpha, IL-1 and lipopolysaccharide in normal fibroblasts, peripheral blood mononuclear cells, synovial cells and chondrocytes. The presence of TSG-6 in synovial fluid suggests a possible role in rheumatoid arthritis. TSG-6 forms a stable complex with components of the serine protease inhibitor, inter-alpha-inhibitor (I alpha I)). TSG-6 potentiates the inhibitory effect of l alpha l on the protease activity of plasmin. Through their cooperative inhibitory effect on plasmin, TSG-6 and l alpha l can modulate the protease network and thus inhibit inflammation.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          Bioinformatics

          Protein Aliases: Hyaluronate-binding protein; TNF alpha-induced protein 6; TNF-stimulated gene 6 protein; TSG-6; Tumor necrosis factor alpha-induced protein 6; tumor necrosis factor alpha-inducible protein 6; tumor necrosis factor induced protein 6; tumor necrosis factor, alpha induced protein 6; tumor necrosis factor, alpha-induced protein 6; Tumor necrosis factor-inducible gene 6 protein; tumor necrosis factor-stimulated gene-6 protein

          View more View less

          Gene Aliases: TNFAIP6; Tnfip6; TSG-6; TSG6

          View more View less

          UniProt ID: (Human) P98066, (Mouse) O08859

          View more View less

          Entrez Gene ID: (Human) 7130, (Mouse) 21930, (Rat) 84397

          View more View less

          Function(s)
          fibronectin binding calcium ion binding protein binding hyaluronic acid binding hydrolase activity metal ion binding carboxylesterase activity molecular_function
          Process(es)
          cell adhesion positive regulation of cell migration negative regulation of inflammatory response ovarian cumulus expansion inflammatory response signal transduction cell-cell signaling hyaluronan metabolic process negative regulation of BMP signaling pathway negative regulation of osteoblast differentiation negative regulation of osteoclast differentiation negative regulation of neutrophil chemotaxis positive regulation of receptor clustering fibronectin fibril organization ovulation
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Argentina flag icon
          Argentina

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-n9r8p:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline