Search
Search
Proteintech
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.
Immunogen sequence: MGGLEPCSRL LLLPLLLAVS GLRPVQAQAQ SDCSCSTVSP GVLAGIVMGD LVLTVLIALA VYFLGRLVPR GRGAAEAATR KQRITETESP YQELQGQRSD VYSDLNTQRP YYK
DAP12 is a transmembrane adaptor protein that is non-covalently associated with several cell surface receptors on natural killer (NK), myeloid, and presumably neuronal cells. DAP12 contains an ITAM domain that preferentially recruits the protein tyrosine kinase Syk to initiate signal transduction. Deficiency of DAP12 results in reduced antigen presentation function by myeloid cells and defects in function of certain NK cell receptors in mice, as well as presenile dementia and bone cysts in humans.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: DNAX adaptor protein 12; DNAX-activation protein 12; KAR-associated protein; killer-activating receptor-associated protein; membrane protein; polycystic lipomembranous osteodysplasia with sclerosing leukoencephalopathy; TYRO protein tyrosine kinase binding protein; TYROBP; unnamed protein product
Gene Aliases: DAP12; KARAP; PLOSL; PLOSL1
UniProt ID: (Human) O43914
Entrez Gene ID: (Human) 7305
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support