Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
Description: This 1TMYS monoclonal antibody reacts with mouse Thymocyte-expressed molecule involved in selection (Themis), also known as Grb2-associating protein (Gasp). Themis is a cytoplasmic protein that is essential for positive selection and the development of single-positive thymocytes. Highest levels of expression are found in CD4+CD8+ double-positive thymocytes, with lower expression found in mature T cells. In addition to Grb1, Themis has been found to associate with PLC gamma 1 and LAT and is quickly phosphorylated by SLP-76 following TCR signaling.
There are 5 isoforms of mouse Themis that are produced as a result of alternative splicing. The 1TMYS antibody recognizes isoforms 1, 2, 3, and 4, but does not recognize isoform 5.
Applications Reported: This 1TMYS antibody has been reported for use in intracellular staining followed by flow cytometric analysis.
Applications Tested: This 1TMYS antibody has been tested by intracellular staining and flow cytometric analysis of mouse thymocytes or splenocytes using the intracellular Fixation & Permeabilization Buffer Set (Product # 88-8824-00) and protocol. Please refer to Best Protocols: Protocol A: Two step protocol for (cytoplasmic) intracellular proteins. This can be used at less than or equal to 0.25 µg per test. A test is defined as the amount (µg) of antibody that will stain a cell sample in a final volume of 100 µL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.
Excitation: 488-561 nm; Emission: 578 nm; Laser: Blue Laser, Green Laser, Yellow-Green Laser.
Filtration: 0.2 µm post-manufacturing filtered.
Themis is a cytoplasmic protein that is essential for positive selection and the development of single-positive thymocytes. Highest levels of expression are found in CD4+CD8+ double-positive thymocytes, with lower expression found in mature T cells. In addition to Grb1, Themis has been found to associate with PLC gamma 1 and LAT and is quickly phosphorylated by SLP-76 following TCR signaling. There are 5 isoforms of mouse Themis that are produced as a result of alternative splicing.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: bA325O24.3; bA325O24.4; FLJ40584; Gasp; Grb2-associating protein; MGC163388; Protein THEMIS; Protein thylex; Thymocyte-expressed molecule involved in selection
Gene Aliases: E430004N04Rik; Gasp; Spot; Themis; thylex; Tsepa
UniProt ID: (Mouse) Q8BGW0
Entrez Gene ID: (Mouse) 210757
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support