Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • Titin Antibodies

          Proteintech

          Titin Polyclonal Antibody, CoraLite® Plus 488

          View all (15) Titin antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers

          Cite Titin Polyclonal Antibody, CoraLite® Plus 488

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (1)
          Titin Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          Titin Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.

          FIGURE: 1 / 1

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          Titin Antibody (CL488-27867) in ICC/IF

          Immunofluorescent analysis of (-20°C Ethanol) fixed NIH/3T3 cells using CoraLite® Plus 488 Titin antibody (CL488-27867) at dilution of 1:200. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          Titin Antibody in Immunocytochemistry (ICC/IF)

          Product Details

          CL488-27867

          Applications
          Tested Dilution
          Publications

          Immunocytochemistry (ICC/IF)

          1:50-1:500
          -
          Product Specifications

          Species Reactivity

          Human, Mouse

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          Titin fusion protein Ag27496

          Conjugate

          CoraLite® Plus 488 CoraLite® Plus 488 CoraLite® Plus 488

          Excitation/Emission Max

          493/522 nm

          Form

          Liquid

          Concentration

          1 mg/mL

          Amount

          100 µg

          Purification

          Antigen affinity chromatography

          Storage buffer

          PBS, pH 7.3, with 50% glycerol, 0.5% BSA

          Contains

          0.05% ProClin 300

          Storage conditions

          -20°C, store in dark

          Shipping conditions

          Wet ice

          Product Specific Information

          Immunogen sequence: MFKSIHEKV SKISETKKSD QKTTESTVTR KTEPKAPEPI SSKPVIVTGL QDTTVSSDSV AKFAVKATGE PRPTAIWTKD GKAITQGGKY KLSEDKGGFF LEIHKTDTSD SGLYTCTVKN SAGSVSSSCK LTIKAIKDTE AQKVSTQKTS EITPQKKAVV QEEISQKALR SEEIKMSEAK SQEKLALKEE ASKVLISEEV KKSAATSLEK SIVHEEITKT SQASEEVRTH AEIK

          Target Information

          Monoclonal Anti-Titin can be used for study of the elastic filaments within sarcomeric structures. It is also useful as a differentiation marker in the separation of rhabdomyosarcomas from other muscle tumors. By indirect immunofluorescence the antibody displays a typical striated staining pattern on frozen sections of chicken skeletal and cardiac muscle tissues. Stains the region of the A-I junction by indirect immunofluorescence. It shows a decoration line 0.05 mm from the end of the A band in electron micro-graphs. In immunoblotting using total extracts of chicken breast muscle the antibody reacts specifically with both bands of the titin doublet, and showed no reaction with nebulin.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: 2.7.11.1; autoimmune antigenic domain; Connectin; DKFZp451N061; FLJ26020; FLJ26409; FLJ32040; FLJ34413; FLJ39564; FLJ43066; Rhabdomyosarcoma antigen MU-RMS-40.14; titan; muscle filament; Titin; ttn; unnamed protein product

          View more View less

          Gene Aliases: 1100001C23Rik; 2310036G12Rik; 2310057K23Rik; 2310074I15Rik; AF006999; AV006427; CMD1G; CMH9; CMPD4; CMYO5; CMYP5; D330041I19Rik; D830007G01Rik; EOMFC; HMERF; L56; LGMD2J; LGMDR10; mdm; MYLK5; SALMY; shru; TMD; TTN

          View more View less

          UniProt ID: (Human) Q8WZ42

          View more View less

          Entrez Gene ID: (Human) 7273, (Mouse) 22138

          View more View less

          Function(s)
          nucleotide binding protease binding protein kinase activity protein serine/threonine kinase activity protein tyrosine kinase activity structural molecule activity calcium ion binding protein binding calmodulin binding ATP binding cytoskeletal protein binding structural constituent of muscle kinase activity transferase activity protein kinase regulator activity enzyme binding protein kinase binding telethonin binding identical protein binding protein homodimerization activity actinin binding metal ion binding actin filament binding muscle alpha-actinin binding structural molecule activity conferring elasticity protein serine kinase activity structural constituent of cytoskeleton protein self-association immunoglobulin receptor superfamily
          Process(es)
          skeletal muscle contraction cardiac muscle hypertrophy muscle contraction striated muscle contraction mitotic chromosome condensation positive regulation of gene expression protein kinase A signaling muscle filament sliding skeletal muscle thin filament assembly skeletal muscle myosin thick filament assembly detection of muscle stretch sarcomere organization sarcomerogenesis positive regulation of protein secretion response to calcium ion cardiac myofibril assembly cardiac muscle tissue morphogenesis cardiac muscle cell development cardiac muscle contraction in utero embryonic development heart morphogenesis adult heart development peptidyl-tyrosine phosphorylation ventricular system development forward locomotion regulation of protein kinase activity cardiac muscle fiber development regulation of catalytic activity striated muscle cell development heart growth
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-d4j86:80/100.66.77.21:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline