Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • UTY Antibodies

          Invitrogen

          UTY Polyclonal Antibody

          View all (5) UTY antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers

          Cite UTY Polyclonal Antibody

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (3)
          UTY Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          UTY Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.

          FIGURE: 1 / 3

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          UTY Antibody (PA5-26082) in ICC/IF

          Immunocytochemistry analysis of UTY in MDA-MB231 cells. Samples were incubated in UTY polyclonal antibody (Product # PA5-26082) followed by Alexa Fluor 488-conjugated goat anti-rabbit lgG (green). Actin filaments have been labeled with Alexa Fluor 555 phalloidin (red). DAPI was used to stain the cell nuclear (blue). {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          UTY Antibody in Immunocytochemistry (ICC/IF)
          UTY Antibody in Western Blot (WB)
          UTY Antibody in Flow Cytometry (Flow)
          UTY Polyclonal Antibody

          Product Details

          PA5-26082

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:1,000
          -

          Immunocytochemistry (ICC/IF)

          1:10-1:50
          -

          Flow Cytometry (Flow)

          1:10-1:50
          -
          Product Specifications

          Species Reactivity

          Human

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          KLH conjugated synthetic peptide between 1247-1276 amino acids from the C-terminal region of human UTY
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          0.5 mg/mL

          Amount

          200 µg

          Purification

          Protein A, Antigen affinity chromatography

          Storage buffer

          PBS, pH 7.4

          Contains

          0.09% sodium azide

          Storage conditions

          Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_2543582

          Target Information

          UTY encodes a protein containing tetratricopeptide repeats which are thought to be involved in protein-protein interactions. This protein is a minor histocompatibility antigen which may induce graft rejection of male stem cell grafts.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: [histone H3]-trimethyl-L-lysine(27) demethylase UTY; Histone demethylase UTY; ubiquitous TPR motif protein UTY; ubiquitously transcribed tetratricopeptide repeat gene, Y chromosome; ubiquitously transcribed tetratricopeptide repeat gene, Y-linked; ubiquitously transcribed TPR gene on Y chromosome; Ubiquitously-transcribed TPR protein on the Y chromosome; Ubiquitously-transcribed Y chromosome tetratricopeptide repeat protein; unnamed protein product; v10; v100; v101; v102; v103; v104; v105; v106; v107; v108; v109; v11; v110; v111; v112; v113; v114; v115; v116; v117; v118; v119; v12; v120; v121; v122; v123; v124; v125; v126; v127; v128; v129; v13; v130; v131; v132; v133; v134; v135; v136; v137; v138; v139; v14; v140; v141; v142; v143; v144; v145; v146; v147; v148; v149; v15; v150; v151; v152; v153; v154; v155; v156; v157; v158; v159; v16; v160; v161; v162; v163; v164; v165; v166; v167; v168; v169; v17; v170; v171; v172; v173; v174; v175; v176; v177; v178; v179; v18; v180; v181; v182; v183; v184; v185; v186; v187; v188; v189; v19; v190; v191; v192; v193; v194; v195; v196; v197; v198; v199; v20; v200; v201; v202; v203; v204; v205; v206; v207; v208; v209; v21; v210; v211; v212; v213; v214; v215; v216; v217; v218; v219; v22; v220; v221; v222; v223; v224; v225; v226; v227; v228; v229; v23; v230; v231; v232; v233; v234; v235; v236; v237; v238; v239; v24; v240; v241; v242; v243; v244; v245; v246; v247; v248; v249; v25; v250; v251; v252; v253; v254; v255; v256; v257; v258; v259; v26; v260; v261; v262; v263; v264; v265; v266; v267; v268; v269; v27; v270; v271; v272; v273; v274; v275; v276; v277; v278; v279; v28; v280; v281; v282; v283; v284; v285; v286; v287; v29; v30; v31; v32; v33; v34; v35; v36; v37; v38; v39; v4; v40; v41; v42; v43; v44; v45; v46; v47; v48; v49; v5; v50; v51; v52; v53; v54; v55; v56; v57; v58; v59; v6; v60; v61; v62; v63; v64; v65; v66; v67; v68; v69; v7; v70; v71; v72; v73; v74; v75; v76; v77; v78; v79; v8; v80; v81; v82; v83; v84; v85; v86; v87; v88; v89; v9; v90; v91; v92; v93; v94; v95; v96; v97; v98; v99

          View more View less

          Gene Aliases: KDM6AL; KDM6C; UTY; UTY1

          View more View less

          UniProt ID: (Human) O14607

          View more View less

          Entrez Gene ID: (Human) 7404

          View more View less

          Function(s)
          RNA polymerase II core promoter proximal region sequence-specific DNA binding oxidoreductase activity chromatin DNA binding histone demethylase activity metal ion binding dioxygenase activity histone demethylase activity (H3-K27 specific)
          Process(es)
          chromatin organization chromatin remodeling heart development regulation of gene expression
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-6b469b8bb7-lcddp:80/100.66.76.150:80.
          git-commit: a334af76dff23450325448aedefe62379591458a
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.48.1-2026.04.16-1.0