Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
還沒有帳戶嗎 ? 立即註冊
  • 產品
    • Antibodies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Greener Products
    • 查看所有產品類別
  • 應用
    • 生物製程
    • 細胞培養和轉染
    • 食品和飲料
    • 實驗室解決方案
    • 製藥和生物製藥
    • 即時定量PCR
    • 半導體分析
    • 臨床和診斷
    • 數位解決方案
    • 分子光譜、同位素與元素分析
    • 查看所有應用與技術
  • 服務
    • 360° CDMO 和 CRO 服務
    • CDMO 服务
    • 临床试验 CRO 服务
    • 客製化服務
    • 金融與租賃服務
    • 儀器服務
    • 實驗室資訊學
    • 授權與商業供應
    • 培訓服務
    • 查看所有服務
  • 支援
    • 線上訂購協助
    • 數位解決方案
    • 產品支持
    • 技術相關資訊
    • 培訓與教育
    • 查看所有協助與支援
  • 熱門商品
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • 我們的服務對象
    • 生物技術
    • 生物製藥
    • CDMO
    • 實驗室診斷
    • 工業與應用科學
  • 特價商品
  • 聯絡我們
  • 快速訂購
  • 訂單狀態與貨物追蹤
  • 文件和證書
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • 訂單查詢
          • 快速訂購
          • 登入
            登入
            還沒有帳戶嗎 ? 立即註冊
            • 帳戶
            • 訂單
            • Connect: Lab, Data, Apps
            • 客製化產品與項目
            • 儀器管理
          • Primary Antibodies ›
          • VDAC Antibodies

          Rockland

          VDAC/Porin Polyclonal Antibody

          View all (39) VDAC antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support

          Cite VDAC/Porin Polyclonal Antibody

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (4)
          VDAC/Porin Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          VDAC/Porin Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.

          FIGURE: 1 / 4

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          VDAC/Porin Antibody (600-101-HB2) in ICC/IF

          Immunofluorescence of Goat Anti-VDAC/Porin Antibody. Cell Line: HeLa Cells. Fixation: 100% MeOH. Permeabilization: 0.3% Triton X-100. Primary Antibody: Anti-VDAC1/Porin at 5µg/mL overnight at 2-8°C. Secondary Antibody: Donkey Anti-Goat IgG DyLight™488 (p/n 605-741-125) at 5µg/mL for 1hr at RT. Nuclear counter stain: DAPI. Location: Mitochondrion, Mitochondrion Outer Membrane, Nucleus. A). DAPI. B). V... View More {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          VDAC/Porin Antibody in Immunocytochemistry (ICC/IF)
          VDAC/Porin Antibody in Western Blot (WB)
          VDAC/Porin Antibody in Western Blot (WB)
          VDAC/Porin Antibody in Western Blot (WB)
          VDAC/Porin Polyclonal Antibody

          Product Details

          600-101-HB2

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:1,000
          -

          Immunocytochemistry (ICC/IF)

          15 µg/mL
          -

          ELISA (ELISA)

          1:10,000-1:50,000
          -
          Product Specifications

          Species Reactivity

          Human, Mouse, Rat

          Host/Isotype

          Goat / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          VDAC/Porin Antibody was prepared from whole goat serum produced by repeated immunizations with a synthetic peptide corresponding to N-terminal portion of Human VDAC1/Porin1.
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          1 mg/mL

          Amount

          100 µg

          Purification

          Affinity chromatography

          Storage buffer

          0.02M potassium phosphate, pH 7.2, with 0.15M NaCl

          Contains

          0.01% sodium azide

          Storage conditions

          -20°C, Avoid Freeze/Thaw Cycles

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          Product Specific Information

          Store vial at -20° C prior to opening. Aliquot contents and freeze at -20° C or below for extended storage. Avoid cycles of freezing and thawing. Centrifuge product if not completely clear after standing at room temperature. This product is stable for several weeks at 4° C as an undiluted liquid. Dilute only prior to immediate use.

          Anti-VDAC/Porin Antibody is directed against human VDAC1/Porin1 protein. A BLAST analysis was used to suggest that this antibody would react with VDAC1/Porin1 including mouse, rat, bovine, pig, and rabbit sources based on 100% homology for the immunogen sequence. Such broad reactivity makes this antibody useful as an excellent loading control.

          Target Information

          Voltage-dependent anion channel (VDAC) proteins are abundant, pore-forming proteins belonging to the eukaryotic mitochondrial porins. It was discovered in the mitochondrial outer membrane. It also is expressed in the plasma membrane. At least three different VDAC genes have been identified in vertebrates. VDAC proteins are known to play an essential role in cellular metabolism and in the early stages of apoptosis. For example, VDAC constitutes a major pathway by which metabolites such as ADP/ATP, succinate and citrate are exchanged between the cytosol and mitochondria. And VDAC1 in the plasma membrane establishes a novel level of apoptosis regulation putatively via its redox activity.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: ARABIDOPSIS THALIANA VOLTAGE DEPENDENT ANION CHANNEL 1; ARABIDOPSIS THALIANA VOLTAGE DEPENDENT ANION CHANNEL 2; ATVDAC1; ATVDAC2; F2G14.210; HSR2; hVDAC1; K9I9.6; K9I9_6; MGC111064; Mitochondrial Porin; mVDAC1; mVDAC5; Non-selective voltage-gated ion channel VDAC1; Outer mitochondrial membrane protein porin 1; Plasmalemmal porin; Porin 31HL; Porin 31HM; PORIN-31-HL; rVDAC1; sperm binding protein 1a; T22N4.9; T22N4_9; unnamed protein product; VD; VDAC-1; VDAC-5; VDAC1; VDAC2; Voltage-dependent anion-selective channel protein 1; Voltage-dependent anion-selective channel protein 5

          View more View less

          Gene Aliases: AL033343; PORIN; VDAC; VDAC-1; VDAC1; Vdac5

          View more View less

          UniProt ID: (Human) P21796, (Mouse) Q60932, (Rat) Q9Z2L0

          View more View less

          Entrez Gene ID: (Human) 7416, (Mouse) 22333, (Rat) 83529

          View more View less

          Function(s)
          nucleotide binding voltage-gated ion channel activity protein binding ATP binding oxysterol binding lipid binding voltage-gated anion channel activity porin activity cholesterol binding protein kinase binding phosphatidylcholine binding identical protein binding ion channel binding ceramide binding anion channel activity protein complex binding voltage-gated ion channel
          Process(es)
          behavioral fear response ion transport anion transport lipid transport apoptotic process chemical synaptic transmission neuron-neuron synaptic transmission learning epithelial cell differentiation calcium ion transmembrane import into mitochondrion positive regulation of apoptotic process negative regulation of apoptotic process transmembrane transport anion transmembrane transport negative regulation of calcium import into the mitochondrion regulation of mitophagy regulation of mitophagy positive regulation of type 2 mitophagy mitochondrial transmembrane transport negative regulation of reactive oxygen species metabolic process transport mitochondrial calcium ion transport regulation of anion transport regulation of anion transmembrane transport ion transmembrane transport regulation of ion transmembrane transport
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          產品訂購 Plus Icon Minus Icon
          • 訂單狀態
          • 訂購協助
          • 快速訂購
          • 現貨供應中心
          • 電子採購
          支持 Plus Icon Minus Icon
          • 協助與支持
          • 聯絡我們
          • 技術支持中心
          • 搜尋文件和證書
          • 回報網站問題
          資源 Plus Icon Minus Icon
          • 學習中心
          • 促銷活動
          • 活動和網路研討會
          • 社群媒體
          關於 Thermo Fisher Plus Icon Minus Icon
          • 關於我們 關於我們
          • 人才招募 人才招募
          • 投資者資訊 投資者資訊
          • 新聞 新聞
          • 企業社會責任 企業社會責任
          • 商標
          • 政策和聲明
          品牌 Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-744bc48644-fvx7n:80/100.66.78.247:80.
          git-commit: 747bde55a712f6f97bf7760408d445eefba4e16f
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.48.0-Offline