FIGURE: 1 / 2
Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.
OSV00006W is specific for VGLUT1 (both isoforms 1 and 2).
Reconstitute with 100 µL of sterile water.
A Na(+)-dependent inorganic phosphate cotransporter; may be involved in neuronal Na+ transport.