Search
Search
Proteintech
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.
Immunogen sequence: MDLDRASLD LMIRLLELEQ DASSAKLLNE KDMNKIKEKI RRLCETVHNK HLDLENITTG HLAMETLLSL TSKRAGDWFK EELRLLGGLD HIVDKVKECV DHLSRDEDEE KLVASLWGAE RCLRVLESVT VHNPENQSYL IAYKDSQLIV SSAKALQHCE ELIQQYNRAE DSICLADSKP LPHQNVTNHV GKAVEDCMRA IIGVLLNLTN DNEWGSTKTG EQDGLIGTAL NCVLQVPKYL PQEQRFDIRV LLFLERERAA QLAESKTDEL IKDAPTTQHD KSGEWQETSG EIQWVSTEKT DGTEEKHKKE EEDEELDLNK ALQHAGKHME DCIVASYTAL LLGCLCQESP INVTTVREYL PEGDFSIMTE MLKKFLSFMN LTCAVGTTGQ KSISRVIEYL EHC (1-402 aa encoded by BC017393)
Transcriptional repressor which may play a role in development of the central nervous system (CNS).
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: dioxin-inducible factor 2; friend of EBNA2; friend of EBNA2 (Epstein-Barr virus nuclear protein 2); friend of EBNA2 protein; KIAA0261; unnamed protein product; wings apart-like homolog
Gene Aliases: A530089A20Rik; DIF-2; FOE; KIAA0261; WAPAL
UniProt ID: (Human) Q7Z5K2, (Mouse) Q65Z40
Entrez Gene ID: (Human) 23063, (Mouse) 218914
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support