Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • WBSCR22 Antibodies

          Abnova

          WBSCR22 Polyclonal Antibody, MaxPab™

          View all (6) WBSCR22 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers

          Cite WBSCR22 Polyclonal Antibody, MaxPab™

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (1)
          WBSCR22 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          WBSCR22 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.

          FIGURE: 1 / 1

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          WBSCR22 Antibody (H00114049-B01P) in WB

          WBSCR22 MaxPab polyclonal antibody. Western Blot analysis of WBSCR22 expression in HeLa S3 NE. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          WBSCR22 Antibody in Western Blot (WB)
          WBSCR22 Polyclonal Antibody, MaxPab™

          Product Details

          H00114049-B01P

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:500-1:1,000
          -

          Immunocytochemistry (ICC/IF)

          10-100 µg/mL
          -
          Product Specifications

          Species Reactivity

          Human

          Host/Isotype

          Mouse / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          WBSCR22 (NP_059998.2, 1 a.a. approximately 281 a.a) full-length human protein.
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          0.84 mg/mL

          Amount

          50 µg

          Purification

          Protein A

          Storage buffer

          PBS, pH 7.4

          Contains

          no preservative

          Storage conditions

          -20°C, Avoid Freeze/Thaw Cycles

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          Product Specific Information

          Product may be used with Western Blot (Cell lysate).

          The Isotype of this product is composed of an IgG Mixture.

          Immunogen sequence: MASRGRRPEH GGPPELFYDE TEARKYVRNS RMIDIQTRMA GRALELLYLP ENKPCYLLDI GCGTGLSGSY LSDEGHYWVG LDISPAMLDE AVDREIEGDL LLGDMGQGIP FKPGTFDGCI SISAVQWLCN ANKKSENPAK RLYCFFASLF SVLVRGSRAV LQLYPENSEQ LELITTQATK AGFSGGMVVD YPNSAKAKKF YLCLFSGPST FIPEGLSENQ DEVEPRESVF TNERFPLRMS RRGMVRKSRA WVLEKKERHR RQGREVRPDT QYTGRKRKPR F

          Target Information

          The WBSCR22 gene, also known as Williams-Beuren Syndrome Chromosome Region 22, encodes a protein that participates in the biogenesis of the 40S ribosomal subunit. This gene is one of the 26 deleted in Williams-Beuren syndrome, which is characterized by cognitive delays, congenital heart and vascular diseases, and distinct facial features. WBSCR22 features a nuclear localization signal and an S-adenosylmethionine (SAM) binding motif typical of methyltransferases. The protein enhances cell survival and proliferation, with increased expression linked to poor prognosis in various cancers, including glioma and breast cancer. WBSCR22 modulates key signaling pathways such as Akt/GSK3B and beta-catenin/CyclinD1, and is a direct target of miR-146b-5p. Moreover, its downregulation suppresses tumor growth, suggesting its potential as a therapeutic target in oncology.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: 18S rRNA (guanine-N(7))-methyltransferase; Bud site selection protein 23 homolog; Metastasis-related methyltransferase 1; methyltransferase; probable 18S rRNA (guanine-N(7))-methyltransferase; ribosome biogenesis methyltransferase WBSCR22; rRNA methyltransferase and ribosome maturation factor; unnamed protein product; Williams-Beuren candidate region putative methyltransferase; Williams-Beuren syndrome chromosomal region 22 protein; Williams-Beuren syndrome chromosome region 22

          View more View less

          Gene Aliases: BUD23; HASJ4442; HUSSY-03; HUSSY-3; MERM1; PP3381; WBMT; WBSCR22

          View more View less

          UniProt ID: (Human) O43709

          View more View less

          Entrez Gene ID: (Human) 114049

          View more View less

          Function(s)
          RNA binding protein binding methyltransferase activity S-adenosylmethionine-dependent methyltransferase activity rRNA (guanine) methyltransferase activity transferase activity protein heterodimerization activity
          Process(es)
          chromatin organization rRNA processing methylation ribosome biogenesis rRNA (guanine-N7)-methylation positive regulation of rRNA processing
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-6b469b8bb7-b97mm:80/100.66.72.101:80.
          git-commit: a334af76dff23450325448aedefe62379591458a
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.48.1-2026.04.16-1.0