Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • WNT11 Antibodies

          Invitrogen

          WNT11 Polyclonal Antibody

          View all (9) WNT11 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers

          Cite WNT11 Polyclonal Antibody

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (1)
          WNT11 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Group 53 Created with Sketch.
          WNT11 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Group 53 Created with Sketch.

          FIGURE: 1 / 1

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          WNT11 Antibody (PA5-34317) in IHC (P)

          Immunohistochemistry analysis of WNT11 in human liver. Samples were incubated with WNT11 polyclonal antibody (Product # PA5-34317). Formalin-fixed, paraffin-embedded tissue after heat-induced antigen retrieval. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          WNT11 Antibody in Immunohistochemistry (Paraffin) (IHC (P))

          Product Details

          PA5-34317

          Applications
          Tested Dilution
          Publications

          Immunohistochemistry (Paraffin) (IHC (P))

          10 µg/mL
          -
          Product Specifications

          Species Reactivity

          Bat, Bovine, Dog, Chicken, Horse, Hamster, Human, Mouse, Non-human primate, Pig, Rabbit, Rat

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          Synthetic 15 amino acid peptide from internal region of human WNT11.

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          1 mg/mL

          Amount

          50 µg

          Purification

          Antigen affinity chromatography

          Storage buffer

          PBS

          Contains

          0.1% sodium azide

          Storage conditions

          Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_2551669

          Target Information

          Ligand for members of the frizzled family of seven transmembrane receptors. Probable developmental protein. May be a signaling molecule which affects the development of discrete regions of tissues. Is likely to signal over only few cell diameters.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: Protein Wnt-11; secreted glycoprotein; unnamed protein product; wingless-related MMTV integration site 11; wingless-type MMTV integration site family, member 11; Wnt-11 protein; WNT11

          View more View less

          Gene Aliases: HWNT11; WNT-11; WNT11

          View more View less

          UniProt ID: (Chicken) P49339, (Human) O96014, (Mouse) P48615

          View more View less

          Entrez Gene ID: (Rabbit) 100009030, (Bovine) 613288, (Chicken) 395562, (Horse) 100064184, (Pig) 100521912, (Dog) 485183, (Human) 7481, (Rat) 140584, (Mouse) 22411, (Chinese hamster) 100774690

          View more View less

          Function(s)
          GTPase activator activity frizzled binding cytokine activity protein kinase activator activity chemoattractant activity receptor binding protein binding molecular_function transcription regulatory region DNA binding intercellular signal molecule
          Process(es)
          osteoblast differentiation epithelial to mesenchymal transition primary heart field specification secondary heart field specification outflow tract morphogenesis atrial septum development planar cell polarity pathway involved in axis elongation protein phosphorylation negative regulation of cell proliferation positive regulation of gene expression negative regulation of gene expression cell migration neuron differentiation bone mineralization negative regulation of cell growth adrenal gland development positive regulation of cell migration negative regulation of cell migration activation of protein kinase activity positive regulation of transforming growth factor beta2 production protein localization to cell surface embryonic appendage morphogenesis positive regulation of apoptotic process negative regulation of apoptotic process positive regulation of GTPase activity skin morphogenesis cell fate commitment maintenance of epithelial cell apical/basal polarity myoblast differentiation negative regulation of transcription, DNA-templated positive regulation of transcription, DNA-templated paraxial mesoderm formation notochord morphogenesis embryonic viscerocranium morphogenesis artery morphogenesis positive chemotaxis positive regulation of stress fiber assembly convergent extension involved in axis elongation canonical Wnt signaling pathway Wnt signaling pathway, planar cell polarity pathway cloacal septation face morphogenesis ventricular septum morphogenesis lung-associated mesenchyme development negative regulation of cell death ureteric bud morphogenesis planar cell polarity pathway involved in gastrula mediolateral intercalation somite development neuroendocrine cell differentiation secondary palate development bicellular tight junction assembly cellular response to mechanical stimulus mesonephric duct development negative regulation of mesenchymal cell proliferation positive regulation of protein kinase C signaling positive regulation of heart induction by negative regulation of canonical Wnt signaling pathway negative regulation of canonical Wnt signaling pathway negative regulation of fibroblast growth factor production primary palate development kidney development ventricular septum development apoptotic process signal transduction Wnt signaling pathway, calcium modulating pathway mesenchymal cell proliferation Wnt signaling pathway cell differentiation response to nutrient levels intracellular signal transduction non-canonical Wnt signaling pathway embryonic skeletal system development palate development negative regulation of cartilage development cellular response to retinoic acid stem cell proliferation epithelial cell apoptotic process positive regulation of epithelial cell apoptotic process negative regulation of stem cell proliferation positive regulation of protein kinase activity cell-cell signaling multicellular organism development organ morphogenesis
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-6b469b8bb7-hrdnk:80/100.66.78.145:80.
          git-commit: a334af76dff23450325448aedefe62379591458a
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.48.1-2026.04.16-1.0