Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Immunogen sequence: ELFQYFGPQA LGQPQTPLAG PGLRPDKPLE AQLLLNGFHH VGAPARKFPT SAPGSLSPDA HLPPSQLLGS SSDSLPTSPP PDDSLSLKVF RCLVCQAFST DSLELLLYHC SIGRSLPEAE WKEVAGDTHR C
Highest antigen sequence identity to the following orthologs: Mouse - 92%, Rat - 90%.
ZFHX2 gene ontology annotations related to this gene include adult behavior; regulation of sensory perception of pain; regulation of transcription, DNA-templated.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support