Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Immunogen sequence: KSFGRSAHLQ AHQKVHTGDK PYKCDECGKG FKWSLNLDMH QRVHTGEKPY
For short term use, store at 2-8°C up to 1 week. For long term storage, store at -20°C in small aliquots to prevent freeze-thaw cycles.
Predicted homology: Cow: 93%; Dog: 93%; Guinea Pig: 86%; Horse: 93%; Human: 100%; Mouse: 86%; Pig: 93%; Rabbit: 92%; Rat: 93%; Zebrafish: 85%
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: hzfp; Kruppel-associated box protein; unnamed protein product; Zinc finger protein 226
Gene Aliases: ZNF226
UniProt ID: (Human) Q9NYT6
Entrez Gene ID: (Human) 7769
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support