Search
Search
Abnova
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MMQPSLDIKP FMSFPVDSSS AVGLFPNFNT MDPVQKAVIN HTFGVSIPPK KKQVISCNVC QLRFNSDSQA EAHYKGSKHA KKVKALDATK NKPKMVPSKD SAKANPSCSI TPITGNNSDK SEDKGKLKAS SSSQPSSSES GSFLLKSGTT PLPPGAATSP SKSTNGAPGT VVESEEEKAK KLLYCSLCKV AVNSLSQLEA HNTGSKHKTM VEARNGAGPI KSYPRPGSRL KMQNGSKGSG LQNKTFHCEI CDVHVNSEIQ LKQHISSRRH KDRVAGKPLK PKYSPYNKLQ RSPSILAAKL AFQKDMMKPL APAFLSSPLA AAAAVSSALS LPPRPSASLF QAPAIPPALL RPGHGPIRAT PASILFAPY
ZNF385B contains 4 U1-type zinc fingers. ZNF385B is expressed in brain with a potential role in RNA maturation, stability, or transcription regulation. There are 5 known isoforms.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: unnamed protein product; ZFN533; Zinc finger protein 385B; Zinc finger protein 533
Gene Aliases: ZNF385B; ZNF533
UniProt ID: (Human) Q569K4
Entrez Gene ID: (Human) 151126
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support