Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Clinical Genomics
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          • Primary Antibodies ›
          • ZNFX1 Antibodies

          Abnova

          ZNFX1 Polyclonal Antibody, MaxPab™

          View all (3) ZNFX1 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support

          Cite ZNFX1 Polyclonal Antibody, MaxPab™

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (1)
          ZNFX1 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          ZNFX1 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.

          FIGURE: 1 / 1

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          ZNFX1 Antibody (H00057169-B01P) in WB

          Western Blot analysis of ZNFX1 expression in transfected 293T cell line by ZNFX1 MaxPab polyclonal antibody. Lane 1: ZNFX1 transfected lysate(210.98 KDa). Lane 2: Non-transfected lysate. . {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          ZNFX1 Antibody in Western Blot (WB)
          ZNFX1 Polyclonal Antibody, MaxPab™

          Product Details

          H00057169-B01P

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:500-1:1,000
          -

          Immunocytochemistry (ICC/IF)

          10-100 µg/mL
          -
          Product Specifications

          Species Reactivity

          Human

          Host/Isotype

          Mouse / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          ZNFX1 (NP_066363.1, 1 a.a. approximately 1918 a.a) full-length human protein.
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          0.5 mg/mL

          Amount

          50 µg

          Purification

          Protein A

          Storage buffer

          PBS, pH 7.4

          Contains

          no preservative

          Storage conditions

          -20°C, Avoid Freeze/Thaw Cycles

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          Product Specific Information

          Product may be used with Western Blot (Transfected lysate).

          The Isotype of this product is composed of an IgG Mixture.

          Immunogen sequence: MEERRPHLDA RPRNSHTNHR GPVDGELPPR ARNQANNPPA NALRGGASHP GRHPRANNHP AAYWQREERF RAMGRNPHQG RRNQEGHASD EARDQRHDQE NDTRWRNGNQ DCRNRRPPWS NDNFQQWRTP HQKPTEQPQQ AKKLGYKFLE SLLQKDPSEV VITLATSLGL KELLSHSSMK SNFLELICQV LRKACSSKMD RQSVLHVLGI LKNSKFLKVC LPAYVVGMIT EPIPDIRNQY PEHISNIISL LQDLVSVFPA SSVQETSMLV SLLPTSLNAL RASGVDIEEE TEKNLEKVQT IIEHLQEKRR EGTLRVDTYT LVQPEAEDHV ESYRTMPIYP TYNEVHLDER PFLRPNIISG KYDSTAIYLD THFRLLREDF VRPLREGILE LLQSFEDQGL RKRKFDDIRI YFDTRIITPM CSSSGIVYKV QFDTKPLKFV RWQNSKRLLY GSLVCMSKDN FETFLFATVS NREQEDLCRG IVQLCFNEQS QQLLAEVQPS DSFLMVETTA YFEAYRHVLE GLQEVQEEDV PFQRNIVECN SHVKEPRYLL MGGRYDFTPL IENPSATGEF LRNVEGLRHP RINVLDPGQW PSKEALKLDD SQMEALQFAL TRELAIIQGP PGTGKTYVGL KIVQALLTNE SVWQISLQKF PILVVCYTNH ALDQFLEGIY NCQKTSIVRV GGRSNSEILK QFTLRELRNK REFRRNLPMH LRRAYMSIMT QMKESEQELH EGAKTLECTM RGVLREQYLQ KYISPQHWES LMNGPVQDSE WICFQHWKHS MMLEWLGLGV GSFTQSVSPA GPENTAQAEG DEEEEGEEES SLIEIAEEAD LIQADRVIEE EEVVRPQRRK KEESGADQEL AKMLLAMRLD HCGTGTAAGQ EQATGEWQTQ RNQKKKMKKR VKDELRKLNT MTAAEANEIE DVWQLDLSSR WQLYRLWLQL YQADTRRKIL SYERQYRTSA ERMAELRLQE DLHILKDAQV VGMTTTGAAK YRQILQKVEP RIVIVEEAAE VLEAHTIATL SKACQHLILI GDHQQLRPSA NVYDLAKNFN LEVSLFERLV KVNIPFVRLN YQHRMCPEIA RLLTPHIYQD LENHPSVLKY EKIKGVSSNL FFVEHNFPEQ EIQEGKSHQN QHEAHFVVEL CKYFLCQEYL PSQITILTTY TGQLFCLRKL MPAKTFAGVR VHVVDKYQGE ENDIILLSLV RSNQEGKVGF LQISNRICVA LSRAKKGMYC IGNMQMLAKV PLWSKIIHTL RENNQIGPML RLCCQNHPET HTLVSKASDF QKVPEGGCSL PCEFRLGCGH VCTRACHPYD SSHKEFQCMK PCQKVICQEG HRCPLVCFQE CQPCQVKVPK TIPRCGHEQM VPCSVPESDF CCQEPCSKSL RCGHRCSHPC GEDCVQLCSE MVTIKLKCGH SQPVKCGHVE GLLYGGLLVK CTTKCGTILD CGHPCPGSCH SCFEGRFHER CQQPCKRLLI CSHKCQEPCI GECPPCQRTC QNRCVHSQCK KKCGELCSPC VEPCVWRCQH YQCTKLCSEP CNRPPCYVPC TKLLVCGHPC IGLCGEPCPK KCRICHMDEV TQIFFGFEDE PDARFVQLED CSHIFEVQAL DRYMNEQKDD EVAIRLKVCP ICQVPIRKNL RYGTSIKQRL EEIEIIKEKI QGSAGEIATS QERLKALLER KSLLHQLLPE DFLMLKEKLA QKNLSVKDLG LVENYISFYD HLASLWDSLK KMHVLEEKRV RTRLEQVHEW LAKKRLSFTS QELSDLRSEI QRLTYLVNLL TRYKIAEKKV KDSIAVEVYS VQNILEKTCK FTQEDEQLVQ EKMEALKATL PCSGLGISEE ERVQIVSAIG YPRGHWFKCR NGHIYVIGDC GGAMERGTCP DCKEVIGGTN HTLERSNQLA SEMDGAQHAA WSDTANNLMN FEEIQGMM

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: NFX1-type zinc finger-containing protein 1; unnamed protein product

          View more View less

          Gene Aliases: IMD91; KIAA1404; ZNFX1

          View more View less

          UniProt ID: (Human) Q9P2E3

          View more View less

          Entrez Gene ID: (Human) 57169

          View more View less

          Function(s)
          RNA binding helicase activity zinc ion binding metal ion binding
          Process(es)
          activation of innate immune response immune system process chromatin silencing by small RNA defense response to bacterium negative regulation of viral genome replication innate immune response defense response to virus
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Hong Kong flag icon
          Hong Kong

          Your items have has been added!


          Host server : magellan-search-blue-744bc48644-454p7:80/100.66.72.101:80.
          git-commit: 747bde55a712f6f97bf7760408d445eefba4e16f
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.48.0-Offline