Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • beta-Arrestin 2 Antibodies

          Invitrogen

          beta-Arrestin 2 Polyclonal Antibody

          View all (15) beta-Arrestin 2 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers

          Cite beta-Arrestin 2 Polyclonal Antibody

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (3)
          beta-Arrestin 2 Antibody in Immunohistochemistry (IHC)
          Group 53 Created with Sketch.
          beta-Arrestin 2 Antibody in Immunohistochemistry (IHC)
          Group 53 Created with Sketch.

          FIGURE: 1 / 3

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          beta-Arrestin 2 Antibody (PA5-113860) in IHC

          Immunohistochemistry of Human Skin using a beta-Arrestin 2 Polyclonal Antibody (Product # PA5-113860). {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          beta-Arrestin 2 Antibody in Immunohistochemistry (IHC)
          beta-Arrestin 2 Antibody in Western Blot (WB)
          beta-Arrestin 2 Antibody in Western Blot (WB)

          Product Details

          PA5-113860

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:1,000
          -

          Immunohistochemistry (IHC)

          1 mg/mL
          -
          Product Specifications

          Species Reactivity

          Human, Mouse

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          Synthetic peptide directed towards the middle region of human ARRB2

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          0.5 mg/mL

          Amount

          50 µg

          Purification

          Affinity chromatography

          Storage buffer

          PBS with 2% sucrose

          Contains

          0.09% sodium azide

          Storage conditions

          -20°C, Avoid Freeze/Thaw Cycles

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_2884375

          Product Specific Information

          Immunogen sequence: RLVIRKVQFA PEKPGPQPSA ETTRHFLMSD RSLHLEASLD KELYYHGEPL

          For short term use, store at 2-8°C up to 1 week. For long term storage, store at -20°C in small aliquots to prevent freeze-thaw cycles.

          Predicted homology: Cow: 100%; Dog: 100%; Guinea Pig: 86%; Horse: 100%; Human: 100%; Mouse: 93%; Pig: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 93%

          Target Information

          Beta-arrestin-2, or ARRB2, is an adaptor protein belonging to the arrestin family with an N- arrestin domain and a C- arrestin domain. ARRB2 is a regulatory protein involved in heterotrimeric G protein-coupled receptor desensitization and is known to regulate beta2-adrenergic receptor (beta2AR) function by enhancing beta2AR mediated nuclear translocation of ERK. ARRB2 binds to phosphorylated beta2ARs, thereby causing a significant impairment of their capacity to activate G(S) proteins. Along with AIP4, ARRB2 acts as an endosomal sorting molecule that mediates CXCR4 entry into a degradative pathway. It may also be involved in hormone-specific desensitization of TSH receptors. ARRB2 is a Ca2+ binding protein of the retinal outer segments and binds to P-rhodopsin and is also involved in synaptic transmission in photoreceptor cells. It is widely expressed in most of the tissues, especially neuronal tissues and spleen.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: activated GPCR interaction protein; ARRB2; arrestin 3; Arrestin beta-2; beta arr2; Beta-arrestin-2; Non-visual arrestin-3; unnamed protein product

          View more View less

          Gene Aliases: AI326910; ARB2; ARR2; ARRB2; AW122872; BARR2

          View more View less

          UniProt ID: (Human) P32121, (Mouse) Q91YI4

          View more View less

          Entrez Gene ID: (Human) 409, (Mouse) 216869

          View more View less

          Function(s)
          G-protein coupled receptor binding receptor binding protein binding enzyme binding protein binding, bridging ubiquitin protein ligase binding angiotensin receptor binding D1 dopamine receptor binding protein kinase B binding binding, bridging protein domain specific binding alpha-1A adrenergic receptor binding alpha-1B adrenergic receptor binding type 1 angiotensin receptor binding follicle-stimulating hormone receptor binding type 2A serotonin receptor binding platelet activating factor receptor binding protein complex binding protein complex scaffold mitogen-activated protein kinase binding 14-3-3 protein binding scaffold/adaptor protein
          Process(es)
          desensitization of G-protein coupled receptor protein signaling pathway G-protein coupled receptor internalization positive regulation of receptor internalization transcription from RNA polymerase II promoter signal transduction transforming growth factor beta receptor signaling pathway I-kappaB kinase/NF-kappaB signaling adult walking behavior regulation of G-protein coupled receptor protein signaling pathway negative regulation of signal transduction positive regulation of cardiac muscle hypertrophy positive regulation of gene expression protein transport protein ubiquitination sensory perception of pain negative regulation of protein ubiquitination receptor internalization negative regulation of NF-kappaB transcription factor activity positive regulation of synaptic transmission, dopaminergic negative regulation of interleukin-1 beta production negative regulation of interleukin-12 production negative regulation of interleukin-6 production negative regulation of tumor necrosis factor production negative regulation of toll-like receptor signaling pathway negative regulation of I-kappaB kinase/NF-kappaB signaling proteasome-mediated ubiquitin-dependent protein catabolic process negative regulation of natural killer cell mediated cytotoxicity modulation of synaptic transmission positive regulation of protein kinase B signaling negative regulation of protein kinase B signaling excitatory postsynaptic potential cell chemotaxis positive regulation of ERK1 and ERK2 cascade postsynaptic signal transduction beta-arrestin-dependent dopamine receptor signaling pathway positive regulation of intracellular signal transduction positive regulation of cardiac muscle cell differentiation regulation of protein phosphorylation positive regulation of protein phosphorylation desensitization of G-protein coupled receptor protein signaling pathway by arrestin transport endocytosis G-protein coupled receptor signaling pathway positive regulation of protein ubiquitination positive regulation of peptidyl-serine phosphorylation negative regulation of GTPase activity negative regulation of smooth muscle cell apoptotic process follicle-stimulating hormone signaling pathway negative regulation of cysteine-type endopeptidase activity involved in apoptotic process positive regulation of peptidyl-tyrosine phosphorylation detection of temperature stimulus involved in sensory perception of pain positive regulation of calcium ion transport regulation of androgen receptor signaling pathway negative regulation of release of cytochrome c from mitochondria positive regulation of DNA biosynthetic process
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-6b469b8bb7-b97mm:80/100.66.72.101:80.
          git-commit: a334af76dff23450325448aedefe62379591458a
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.48.1-2026.04.16-1.0