Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
FIGURE: 1 / 12
Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.
PA5-16863 targets Tubulin Beta in IHC (P), WB, and IP applications and shows reactivity with Amphibian, Bovine, Chicken, Fungi, Gerbil, Guinea Pig, Human, mouse, Porcine, Rat, Sea Urchin, and Yeast samples.
The PA5-16863 immunogen is a synthetic peptide derived from aa 416-430 of human tubulin beta.
Beta tubulins are one of two core protein families (alpha and beta tubulins) that heterodimerize and assemble to form microtubules. Beta-III tubulin is primarily expressed in neurons and may be involved in neurogenesis, axon guidance and maintenance. Mutations in the beta tubulin gene are the cause of congenital fibrosis of the extraocular muscles type 3. Beta-III tubulin was also detected in Sertoli cells of the testis and transiently in non-neuronal embryonic tissues. Glutamate residues at the C-terminus of beta III tubulin can be glutamylated. The precise function of such modifications is unclear. Tubulin is phosphorylated on Ser-172 by CDK1 during cell cycle progression. Ser-172 phosphorylation inhibits tubulin incorporation into microtubules.Microtubules, the major cytoskeletal elements found in all eukaryotic cells, consist of Tubulin, which is a dimer of two 55 kDa subunits: alpha and Beta. Microtubules play key roles in chromosome segregation in mitosis, intracellular transport, ciliary and flagellar bending, and structural support of the cytoskeleton. Because beta-tubulin is ubiquitously expressed in all eukaryotic cells, it is frequently used as a loading control for assays involving protein detection, such as Western blotting.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: b-Tubulin; beta Ib tubulin; beta tubulin 1; beta-4 tubulin; beta-tubulin; beta-tubulin T beta15 (aa 1-445); betaIII-tubulin; class II beta-tubulin isotype; class IIa beta-tubulin; class IIb beta-tubulin; class III beta-tubulin; class V beta-tubulin; dJ40E16.7; dystonia 4, torsion (autosomal dominant); epididymis secretory sperm binding protein; HSA10p15 beta-tubulin 4Q; HSA18p11 beta-tubulin 4Q pseudogene; M40 antibody; MGC8685; microtubule protein; neuron-specific class III beta-tubulin; similar to beta tubulin; T beta-15; TUBB; tubulin beta; tubulin beta 8 pseudogene 12; tubulin beta class V; tubulin beta MGC4083; tubulin beta-1 chain; tubulin beta-2A chain pseudogene; Tubulin beta-2B chain; tubulin beta-4 chain; tubulin beta-5 chain; Tubulin beta-8 chain-like protein LOC260334; tubulin beta-III; tubulin, beta 1; tubulin, beta 2; tubulin, beta 2A; tubulin, beta 2B; tubulin, beta 3; tubulin, beta 4; tubulin, beta 4 class IVa; tubulin, beta 6; tubulin, beta polypeptide; tubulin, beta polypeptide 2; tubulin, beta polypeptide paralog; tubulin, beta, 5; unnamed protein product; XTP3TPATP1
Gene Aliases: 2310057H16Rik; 2410129E14Rik; 2810484G07Rik; 3200002H15Rik; B130022C14Rik; bA506K6.1; bA631M21.2; beta-4; beta-5; brdp; CDCBM; CDCBM1; CDCBM5; CDCBM6; CDCBM7; CFEOM3; CFEOM3A; CSCSC1; DYT4; FEOM3; FPVEPD; HsT1601; M(beta)1; M(beta)2; M(beta)3; M(beta)4; M(beta)5; M(beta)6; M40; MACTHC1; OK/SW-cl.56; OOMD; OOMD2; OZEMA2; PMGYSA; RGD1305887; RGD1309427; TUBB; TUBB-5; TUBB1; TUBB2; Tubb2b; TUBB4; TUBB5; TUBB8P12
UniProt ID: (Human) P07437, (Human) Q9H4B7, (Human) Q13885, (Human) Q9BVA1, (Human) Q13509, (Human) P04350, (Human) Q9BUF5, (Human) Q3ZCM7, (Human) A6NNZ2, (Mouse) A2AQ07, (Mouse) Q7TMM9, (Rat) P85108, (Rat) Q3KRE8, (Mouse) Q9CWF2, (Mouse) Q9ERD7, (Rat) Q4QRB4, (Mouse) Q9D6F9, (Rat) P69897, (Mouse) P99024, (Mouse) Q922F4
Entrez Gene ID: (Human) 203068, (Human) 81027, (Human) 7280, (Human) 347733, (Human) 10381, (Human) 10382, (Human) 84617, (Human) 347688, (Human) 260334, (Rat) 679312, (Mouse) 545486, (Mouse) 22151, (Rat) 498736, (Rat) 291081, (Mouse) 73710, (Mouse) 22152, (Rat) 246118, (Rat) 29213, (Mouse) 22153, (Rat) 29214, (Mouse) 22154, (Rat) 307351, (Mouse) 67951
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support