Thermo Scientific Indiko Ultra
Performance that fits your laboratory—today and tomorrow
Why Indiko Ultra?
Performance that fits
Indiko Ultra benchtop analyzers
Indiko Ultra analyzers are designed to adapt to your workflow, scale with demand, and deliver consistent performance—reducing operational disruption.
Everyday reliability
Indiko Ultra analyzers combine analytical credibility, operational clarity, advanced data
protection, and scalable capacity in one dependable clinical chemistry analyzer portfolio.
Automated startup, reflex testing, and dilution workflows reduce manual intervention and streamline routine tasks.
Load samples and reagents at any time and run tests in any order—supporting continuous, uninterrupted workflows.
Simultaneously run multiple assays, including sodium, potassium, and chloride, with direct ISE technology for accurate, dilution-free measurements.
  • Up to 200 photometric tests/hour (Indiko Ultra)
  • Up to 350 photometric tests/hour (Indiko Ultra XL)
  • Up to 300,000 tests per year
Low reagent consumption, minimal water usage (1.5–2.5 L/hour), and disposable cuvettes reduce operating costs.
Enhanced cybersecurity, LIS connectivity, and full traceability support confident, compliant laboratory operations.
High performance Indiko reagents
Thermo Scientific™ system reagents for Indiko™ analyzers bring high quality methodologies and smooth operations to your lab.
The extensive test menu features organ, body, and lifestyle panels.
The assays are designed for ease of use thanks to validation and compliance as well as barcoding.
Our partnership brings to you 
Extended menu. Assays from Sentinel Diagnostics expand the Indiko analyzer menu, supporting a broader clinical chemistry reagent portfolio, including the FOB Gold NG Wide reagent for gastrointestinal screening. 
Validated kits. The assays have ready-to-use system applications and they are validated for the Indiko analyzers, to support quick, accurate, and reliable results.
Get a closer look at how Indiko analyzers simplify your workflow. Discover a broad test menu and validated, barcoded assays designed for smooth, reliable performance—download the brochure to learn more.
Designed for efficiency across every step of your workflow
Flexible sample and reagent handling
  • Sample volumes from 2–120 µL across multiple tube and cup formats
  • Automatic barcode identification for samples and reagents
  • Real-time reagent tracking with alerts for low volume, expiration, and lot traceability
Advanced automation and workflow control
  • Continuous sample loading with walk-away time up to 3 hours
  • Automatic reruns and clot detection via pressure sensors
  • Reflex testing based on predefined result thresholds
Robust data and quality management
  • Comprehensive data handling with LIS integration and customizable reporting formats
  • Built-in QC tools with Westgard rules and real-time performance monitoring
  • Easy cost-per-test tracking and operational insights
Results you can stand behind
  • Photometric measurement based on Beer-Lambert principles for precise quantification
  • Stable temperature control at 37°C for consistent reactions
  • TENCELL cuvette technology supports contamination-free, low-volume analysis
Cybersecurity compliance
Indiko Ultra analyzers include an updated cybersecurity architecture aligned with current standards.
System protection
  • Hardened operating system to help prevent installing any third-party software
  • Kiosk mode to help prevent access to the underlying operating system
Access control
  • Secure login to help prevent unauthorized access
  • Restricted user privileges based on role
  • Certificate-based authentication to enable secure access for trained service engineers
Data protection
  • Encrypted database backups and data exports
  • Secure data decommissioning and media sanitization to support protected data handling when the system is taken out of service
These enhancements are designed to support compliance with current and upcoming cybersecurity and regulatory requirements and facilitate integration into modern IT environments. Enhanced cybersecurity features are designed to reduce the attack surface compared to general-purpose operating systems.
Excellent cost efficiency
Global service responsibility
Indiko Ultra analyzers are supported by established service processes designed to ensure system reliability and operational continuity. Thermo Fisher Scientific is responsible for the training and qualification of both its own service engineers and authorized distributor technical personnel who support these analyzers.
Confidence backed by global experience delivered locally
Performance, quality, and insight—working as one
From reliable testing to streamlined quality control and real-time data visibility, this integrated solution helps your lab operate more efficiently while maintaining confidence in every result.
Hear from our team
© 2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.