Search Thermo Fisher Scientific
The custom option offers you the flexibility you need to design your own panels using our 1.3 million TaqMan® Gene Expression Assays, 4.5 million predesigned genotyping assays—including 2,700 inventoried drug-metabolizing enzyme (DME) assays—or your own custom TaqMan® assays. Custom OpenArray® plates include a suite of 11 formats to address your sample throughput needs (see Table 1).
Table 1: Custom OpenArray™ plate format
Custom OpenArray® plates for the QuantStudio® 12K Flex system are offered in 11 formats to provide flexible solutions for configuring your plates for gene expression, miRNA, or genotyping analysis.
Plate format for gene expression & miRNA analysis | # of samples per plate | Minimum order (10 pack) | Total # of samples per minimum order | |
---|---|---|---|---|
Inventoried assays only | Inventoried, made-to-order, and/or custom assays | |||
18 (3X) assays | 48 | 1 | 1 | 480 |
56 assays | 48 | 1 | 1 | 480 |
112 assays | 24 | 1 | 2 | 240 (inv only)/ 480 (inv/custom) |
168 assays | 16 | 1 | 3 | 160 (inv only)/ 480 (inv/custom) |
224 assays | 12 | 1 | 4 | 120 (inv only)/ 480 (inv/custom) |
Note: There are two sets of minimum order requirements for the gene expression offerings. “Inventoried” refers to OpenArray® configurations where all selected assays are inventoried . “Inventoried/made to order/custom” refers to configurations that contain at least one custom or made-to-order assay.
Plate format for genotyping analysis | # of samples per plate | Minimum order (10 pack) | Total # of samples per minimum order |
---|---|---|---|
16 assays | 144 | 1 | 1,440 |
32 assays | 96 | 1 | 960 |
64 assays | 48 | 2 | 960 |
128 assays | 24 | 4 | 960 |
192 assays | 16 | 6 | 960 |
256 assays | 12 | 8 | 960 |
OpenArray® flexible-content panels are pre-configured plates of TaqMan® Gene Expression Assays or TaqMan® SNP Genotyping Assays for a specific pathway, disease, or research focus area. The assays are auto-populated into the custom design tool and the panel can be modified to include or exclude specific assays. This flexible format enables more precise control and simplified assay setup.
Choose an OpenArray® flexible-content panel
OpenArray® fixed-content pathway panels are pre-configured plates of TaqMan® Gene Expression or SNP genotyping assays for a specific biological process, pathway, or disease state.
Pathway panel | Description | ![]() For use on |
---|---|---|
TaqMan® OpenArray® Genetic Barcode Plate 32A | High-throughput tracking and sample identity confirmation; contains a subset of 32 assays specific to genetic genotyping barcodes | 4475386 |
TaqMan® OpenArray® Genetic Barcode Plate 32B | High-throughput tracking and sample identity confirmation; contains a second subset of 32 assays specific to genetic genotyping barcodes | 4475387 |
TaqMan® OpenArray® Genetic Barcode Plate 64 | High-throughput tracking and sample identity confirmation; contains a full set of 64 assays specific to genetic genotyping barcodes | 4475394 |
TaqMan® OpenArray® Human Kinome Plate | Identify profiles of genes involved in the regulation of cellular pathways, contains 828 assays specific to kinase and kinase-related genes | 4475388 |
TaqMan® OpenArray® Human Inflammation Plate | Gene expression markers to characterize key pathways of inflammation response; contains 607 assays specific to human inflammation genes | 4475389 |
TaqMan® OpenArray® Human Stem Cell Plate | Gene expression markers to characterize human embryonic stem cell (hESC) identity; contains 631 assays specific to stem cell–related genes | 4475390 |
TaqMan® OpenArray® Human Cancer Plate | Identify differentially expressed genes involved in key pathways of cancer; contains 648 assays specific to cancer | 4475391 |
TaqMan® OpenArray® Human Signal Transduction Plate | Identify major genes involved in key signaling pathways; contains 597 assays specific to signal transduction–related genes | 4475392 |
TaqMan® OpenArray® Mouse Inflammation Plate | Gene expression markers to characterize key pathways of inflammation response; contains 648 assays specific to mouse inflammation genes | 4475393 |
TaqMan® OpenArray® Pharmacogenomics Panel | Validation of drug-metabolizing enzyme (DME) markers; contains 158 DME assays associated with the PharmaADME Core Marker List | 4475395 |
TaqMan® OpenArray® Pharmacogenomics Express Panel | Validation of drug-metabolizing enzyme (DME) markers; contains 60 DME assays commonly used in pharmacogenomics research | 4488847 |
TaqMan® OpenArray® fixed-content pathway panels are available for use on QuantStudio™ 12K Flex Real-Time PCR System and OpenArray® Real-Time PCR System. However, panels are not compatible with both systems. Please select the correct panel for the system that you own.
For Research Use Only. Not for use in diagnostic procedures.
The custom option offers you the flexibility you need to design your own panels using our 1.3 million TaqMan® Gene Expression Assays, 4.5 million predesigned genotyping assays—including 2,700 inventoried drug-metabolizing enzyme (DME) assays—or your own custom TaqMan® assays. Custom OpenArray® plates include a suite of 11 formats to address your sample throughput needs (see Table 1).
Table 1: Custom OpenArray™ plate format
Custom OpenArray® plates for the QuantStudio® 12K Flex system are offered in 11 formats to provide flexible solutions for configuring your plates for gene expression, miRNA, or genotyping analysis.
Plate format for gene expression & miRNA analysis | # of samples per plate | Minimum order (10 pack) | Total # of samples per minimum order | |
---|---|---|---|---|
Inventoried assays only | Inventoried, made-to-order, and/or custom assays | |||
18 (3X) assays | 48 | 1 | 1 | 480 |
56 assays | 48 | 1 | 1 | 480 |
112 assays | 24 | 1 | 2 | 240 (inv only)/ 480 (inv/custom) |
168 assays | 16 | 1 | 3 | 160 (inv only)/ 480 (inv/custom) |
224 assays | 12 | 1 | 4 | 120 (inv only)/ 480 (inv/custom) |
Note: There are two sets of minimum order requirements for the gene expression offerings. “Inventoried” refers to OpenArray® configurations where all selected assays are inventoried . “Inventoried/made to order/custom” refers to configurations that contain at least one custom or made-to-order assay.
Plate format for genotyping analysis | # of samples per plate | Minimum order (10 pack) | Total # of samples per minimum order |
---|---|---|---|
16 assays | 144 | 1 | 1,440 |
32 assays | 96 | 1 | 960 |
64 assays | 48 | 2 | 960 |
128 assays | 24 | 4 | 960 |
192 assays | 16 | 6 | 960 |
256 assays | 12 | 8 | 960 |