Charging effects in X-ray photoelectron spectroscopy
When X-ray photons strike a surface, they cause the emission of electrons. This, of course, is the basis of the XPS technique. If the surface is electrically insulating, then the emission of electrons causes a positive charge to accumulate at the surface. When a non-monochromated source is used for XPS analysis there are usually a sufficient number of electrons in the region of the sample to limit the effect of the charging. However, when using a monochromated X-ray source for XPS measurements, there are too few stray electrons to control the charging and the resulting positive charge severely affects the XPS spectrum. It causes the peaks in the spectrum to shift to high binding energies and become distorted. For this reason, it is necessary to neutralize the charge on the surface by replenishing electrons from an external source. This stabilizes and controls the charging to within a few electron volts of the neutral state. Any remaining small shift can be removed after data collection is complete, by using the spectrum processing tools contained within the Thermo Scientific Avantage data system.
Combination electron beam and ion beam gun
The use of a single gun to produce both the focused low energy electron beam and the large area ion flux has a number of distinct advantages over the use of independent sources. A new, combined ion/electron gun has been designed which incorporates all the above advantages. This novel design uses a LaB6 emitter to produce a bright source of low energy electrons, with an energy spread of only about 0.3 eV. These are focused to a small spot on the sample. Precise alignment to the X-ray beam is provided using integral X-Y deflectors. Focusing and alignment of this low energy beam is enhanced by the extremely low residual magnetic fields present in the mu-metal analysis chambers.
Features of Charge Compensation for the ESCALAB Xi+ spectrometer
  • Patented ‘in the lens’ multi-mode electrostatic flood source
  • 0 - 5 eV (mode 1) charge compensation
  • 0 - 1000 eV (mode 2) REELS / imaging / alignment
  • Up to 500 µA electron emission
  • Additional dual beam (ion / electron) flood source
Features of Charge Compensation for Nexsa and K-Alpha spectrometers
  • Patented dual beam source
  • Single mode flood gun for all sample types
  • Single mode for all X-ray spot sizes
  • Ultra-low energy beam results in minimal shift from reference positions
  • Flood source assembly incorporating electrostatic deflection for precise alignment
  • REELS capability available for Nexsa
Charging Effects in XPS
Products
Style Sheet to change H2 style to p with em-h2-header class
Style Sheet for Instrument Cards Original
Nexsa G2 XPS
  • Micro-focus X-ray sources
  • Unique multi-technique options
  • Dual-mode ion source for monoatomic & cluster ion depth profiling
K-Alpha XPS
  • High resolution XPS
  • Fast, efficient, automated workflow
  • Ion source for depth profiling
ESCALAB QXi XPS
  • High spectral resolution
  • Multi-technique surface analysis
  • Extensive sample preparation and expansion options
Contact us