Refractive Index Detectors
  • HPLC Detectors
1 - 2 of 2 Products
RefractoMax 521 Refractive Index Detector
For cost-effective analysis of compounds with little or no absorption in the UV range, use the Thermo Scientific™ RefractoMax 521 Refractive Index Detector. It is ideal for detection of alcohols, sugars, carbohydrates, fatty acids and polymers in combination with an isocratic Thermo™ Scientific™...
Vanquish™ Refractive Index Detector Green features
Experience the ease of using the Thermo Scientific™ Vanquish™ Refractive Index Detector (RID) for routine isocratic analysis of substances that lack UV chromophores, such as carbohydrates, lipids, and polymers.
spinner