Transfection Protocols
Our Lipofectamine Reagent Protocols have been optimized for efficiency, viability, and reproducibility across a broad range of cell types. This is often the best place to start, especially in a new cell line. If you find this doesn’t work for your specific cell type, then you can you look to our cell specific protocols for further optimization.
Transfection protocols
No records were found matching your criteria
Cell type | Transfected nucleic acid | Transfection reagent |
---|---|---|
A549 human lung epithelial carcinoma cells | plasmid DNA | Lipofectamine 3000 |
A549 human lung epithelial carcinoma cells | plasmid DNA | Lipofectamine LTX |
ACHN human renal adenocarcinoma cells | plasmid DNA | Lipofectamine LTX |
C6 rat glioma cells | plasmid DNA | Lipofectamine LTX |
Caco-2 human adenocarcinoma cells | plasmid DNA | Lipofectamine LTX |
CHO-K1 cells | plasmid DNA | Lipofectamine LTX |
COS-7 monkey kidney fibroblasts | plasmid DNA | Lipofectamine LTX |
DU 145 human prostate epithelial carcinoma cells | plasmid DNA | Lipofectamine 3000 |
eukaryotic cells | DNA, RNA | DMRIE-C |
eukaryotic cells | plasmid DNA | FreeStyle MAX |
eukaryotic cells | DNA | Lipofectamine |
eukaryotic cells | DNA, RNA | Lipofectamine 2000 |
eukaryotic cells | DNA | Lipofectamine 2000 CD |
eukaryotic cells | DNA | Lipofectamine LTX and PLUS |
eukaryotic cells | siRNA, oligos | Oligofectamine |
eukaryotic cells | pre-transfection DNA complexes | PLUS pre-transfection enhancer |
FreeStyle 293-F cells | plasmid DNA | 293fectin |
GripTite 293 MSR HECs | plasmid DNA | Lipofectamine LTX |
HCT 116 human colorectal carcinoma cells | plasmid DNA | Lipofectamine LTX |
HEK293 cells | plasmid DNA | Lipofectamine LTX |
HEK293 suspension cells | plasmid DNA | 293fectin |
HeLa S3 human cervical adenocarcinoma cells | plasmid DNA | Lipofectamine LTX |
Hep 3B2.1-7 human hepatocellular carcinoma cells | plasmid DNA | Lipofectamine 3000 |
Hep G2 human hepatocellular carcinoma cells | plasmid DNA | Lipofectamine 3000 |
Hep G2 human hepatocellular carcinoma cells | plasmid DNA | Lipofectamine LTX |
Hepa 1-6 mouse hepatoma cells | plasmid DNA | Lipofectamine 3000 |
HL-60 human lymphoblast cells | plasmid DNA | Lipofectamine LTX |
Hs 578T human mammary epithelial carcinoma cells | plasmid DNA | Lipofectamine 3000 |
HT 29 human colorectal adenocarcinoma cells | plasmid DNA | Lipofectamine LTX |
HT-1080 human fibrosarcoma cells | plasmid DNA | Lipofectamine LTX |
HUVEC | plasmid DNA | Lipofectamine LTX |
HUVEC | siRNA, Stealth RNAi | RNAiMAX |
IMR-90 human embryonic lung cells | plasmid DNA | Lipofectamine LTX |
Jurkat human T cell leukemia cells | plasmid DNA | Lipofectamine LTX |
K-562 human myelogenous leukemia cells | plasmid DNA | Lipofectamine LTX |
LNCaP human prostate epithelial carcinoma cells | plasmid DNA | Lipofectamine 3000 |
LNCaP human prostate epithelial carcinoma cells | plasmid DNA | Lipofectamine LTX |
mammalian cells | dsRNA oligo (BLOCK-iT fluorescent oligo) | cationic lipid |
mammalian cells | siRNA, Stealth RNAi + plasmid DNA (co-transfection) | Lipofectamine 2000 |
mammalian cells | siRNA, Stealth RNAi | Lipofectamine 2000 |
mammalian cells | DNA, RNA, oligos | Lipofectin |
mammalian cells | siRNA, Stealth RNAi | RNAiMAX |
mammalian cells, reverse transfection | siRNA, Stealth RNAi | Lipofectamine 2000 |
mammalian cells, reverse transfection | siRNA, Stealth RNAi | RNAiMAX |
MCF 10A human mammary epithelial cells | plasmid DNA | Lipofectamine 3000 |
MCF7 human breast adenocarcinoma cells | plasmid DNA | Lipofectamine 3000 |
MCF7 human breast adenocarcinoma cells | plasmid DNA | Lipofectamine LTX |
MDA-MB-231 human breast adenocarcinoma cells | plasmid DNA | Lipofectamine 3000 |
MDA-MB-231 human breast adenocarcinoma cells | plasmid DNA | Lipofectamine LTX |
MDA-MB-435 cells | plasmid DNA | Lipofectamine LTX |
MDA-MB-453 human breast adenocarcinoma cells | plasmid DNA | Lipofectamine 3000 |
MDA-MB-468 human breast adenocarcinoma cells | plasmid DNA | Lipofectamine 3000 |
MOLT-4 human T cell lymphoblast cells | plasmid DNA | Lipofectamine LTX |
NCI-H23 non-small cell lung cancer cells | plasmid DNA | Lipofectamine 3000 |
NCI-H460 human large cell lung cancer cells | plasmid DNA | Lipofectamine 3000 |
NCI-H460 human large cell lung cancer cells | plasmid DNA | Lipofectamine LTX |
NIH/3T3 mouse embryonic fibroblasts | plasmid DNA | Lipofectamine LTX |
Normal human foreskin fibroblasts (NHFF) | plasmid DNA | Lipofectamine LTX |
NTERA-2 human embryonal carcinoma stem cells | plasmid DNA | Lipofectamine LTX |
PC-12 rat neuronal cells | plasmid DNA | Lipofectamine LTX |
PC-3 human prostate adenocarcinoma cells | plasmid DNA | Lipofectamine LTX |
plant protoplasts | DNA | Lipofectin |
Primary mouse neural progenitor cells (mNPC) | plasmid DNA | Lipofectamine LTX |
RAW 264.7 mouse macrophage cells | plasmid DNA | Lipofectamine LTX |
Sf9, Sf21 | baculovirus DNA | Cellfectin II |
SK-BR-3 human breast adenocarcinoma cells | plasmid DNA | Lipofectamine 3000 |
SK-BR-3 human breast adenocarcinoma cells | plasmid DNA | Lipofectamine LTX |
SK-MEL-28 human malignant melanoma cells | plasmid DNA | Lipofectamine LTX |
SK-OV-3 human ovarian adenocarcinoma cells | plasmid DNA | Lipofectamine LTX |
stem cells, (hMSCs) | siRNA, Stealth RNAi | Lipofectamine 2000 |
stem cells, (hPSCs) in Essential 8 Medium | plasmid DNA, mRNA, RNP | Lipofectamine Stem |
stem cells, (hPSCs) in mTeSR1 Medium on Geltrex matrix | plasmid DNA, mRNA, RNP | Lipofectamine Stem |
stem cells, (hPSCs) in StemFlex Medium | plasmid DNA, mRNA, RNP | Lipofectamine Stem |
stem cells, neural | plasmid DNA, mRNA, RNP | Lipofectamine Stem |
THP-1 human monocytic leukemia cells | plasmid DNA | Lipofectamine LTX |