Search
These stability chambers provide precise control over a wide range of temperature and humidity settings. They are ideal for drug stability studies, shelf-life testing for packaged products, and insect hatching and other large-scale biological research.
Temperature
Control range of 0°C to 60°C (32°F to 140°F) in 0.1°C increments.
Humidity
Control range of above ambient to 95% @ 37°C (98.6°F)
Extensive cabinet options
Doors, shelves, access ports, sockets, shelving, chart recorders and shaker systems.
Extensive humidity system options
Designed to accommodate labs with a variety of water access and drainage needs.
Achieves regulatory compliance
Meets International Conference on Harmonization (ICH) guidelines for drug substance and product storage testing.
|
Large Stability Chamber |
Small Stability Chamber |
Size |
29 cu. ft. |
11 cu. ft. |
Exterior (W x H x D) |
965 x 2248 x 813mm |
965 x 1308 x 813mm |
Interior (W x H x D) |
787 x 1524 x 686mm |
787 x 609 x 686mm |
Net weight |
347kg / 765 lbs |
261kg / 575 lbs |
Shipping weight |
472kg / 1040 lbs |
325kg / 715 lbs |
Temperature |
||
Temperature range |
0°C to 60°C (32°F to 140°F) |
|
Controller, sensor |
Watlow Microprocessor - PID, with RTD sensor |
|
Display |
Digital dual LED, 0.1°C steps readability & setability |
|
Programmability |
Single set point, ramp & soak (to 40 steps) |
|
Temp. uniformity @ 20 to 37°C |
≤ ±0.3°C |
|
Temp. stability @ 20 to 37°C |
≤ ±0.1°C |
|
Humidity |
||
Water consumption / specification |
Max. 12L per day / 3 gal per day; |
|
Relative humidity range |
Above ambient to 95% @ 37°C (98.6°F) ±5.0% RH |
|
Water connection* |
Fill port: 1⁄8" female NPT, drain port: 3⁄8" FPT & 3⁄8" O.D. P-trap right rear, initial fill approx. 1L / 1qt. |
|
Electrical |
||
Voltage |
208-220V, 60Hz |
|
Plug |
NEMA 14-20P |
|
Power switch |
2-Pole |
|
Dry Contacts |
Common, NO, NC |
|
Data Output |
4-20 milliamp, temperature and RH |
|
Access port |
54mm / 2.1in diameter; left side; with exterior and interior silicone stopper |
|
Options |
||
Cabinet Options Available |
Choice of access ports Duplex sockets Choice of chart recorders Infrared CO2 Control Package Choice of door types and styles, including Lexan(tm) inner doors Choice of shaker support systems Temperature mapping and calibration certificate
|
|
Humidity Systems Options Available |
Heatless dryer Condensate pump kit Condensate evaporator Carboy kit
|
|
Order Number |
||