Ion scattering spectroscopy
Ion scattering spectroscopy (ISS) is a technique in which a beam of ions is scattered by a surface. The kinetic energy of scattered ions is measured; peaks are observed corresponding to elastic scattering of ions from atoms at the surface of the sample. Each element at the sample surface produces a peak at a different measured kinetic energy, caused by the momentum transfer between the incident ion and atom. The scattered ion and the scattering atom are normally of different masses, but the total momentum of the atom and ion is conserved. Therefore, as the initially "stationary" atom recoils, some kinetic energy is lost from the scattered ion, and the quantity of lost energy depends on the relative masses of the atom and ion.
Ion scattering data
Experimental geometry for ion scattering spectroscopy.
Experimental geometry for ion scattering spectroscopy.
An ion scattering spectrum typically contains one peak for each element in the sample, with their separation in energy relating to the relative atomic masses of those elements. In some cases, different isotopes of the same element can be resolved, although this usually requires a primary ion heavier than He+. Strong scattering signals are limited to the topmost layer of atoms, so ISS is extremely surface-sensitive. Therefore, the sample must be clean, and even a small quantity of surface contaminant can significantly influence the ion scattering spectrum.
The absolute kinetic energy (ES) of each peak also depends on the energy of the scattered ion beam (E0) because ISS is an energy-loss technique. The relevant quantity when dealing with ISS is the ratio of the scattered and incident ion energies, E/E0. The angle through which the ions are scattered (θ) is also important. Where the ion source and detector are fixed, the angle is a parameter that must be included in the calculations.
Join-the-conversation-banner-in-blogs-social-graphic-1143250-1000x172
Ion scattering energy
The equation shows how the energy of a scattered ion peak is related to the other relevant factors.
ES = Kinetic energy of the scattered ion
M1 = Relative atomic mass of the scattered ion
E0 = Kinetic energy of the primary ion beam
M2 = Relative atomic mass of the scattering surface atom
θ = Scattering angle
For a given instrument, θ is normally a constant (it is ~130° on the Thermo Scientific ESCALAB Xi+ instrument, because the angle between the ion gun and the input lens is 50°). M1 is constant for a given source gas, which is usually He, but sometimes Ne, Ar and other inert gases are used. E0 is normally constant in a given experiment. Therefore, this equation can be used to determine the mass of a scattering atom, M2, from its peak position in the spectrum or to predict the position of a peak for a given atom.
whatisxps_ion-02
Sometimes, E0 is unknown at the start of an experiment. The solution is to calibrate the energy using a standard sample, preferably of a pure, metallic element for which M2 is known, such as gold. Once the scattered peak energy from the standard sample is measured, the only remaining unknown is E0, which can be calculated. Once E0 is known, the main aim of ISS is usually to associate scattered peaks of energy ES with atoms of mass M2. The equation may be simplified to a quadratic equation in (M2/M1).
Let M = (M2/M1), E = (ES/E0), C = cos θ, S = sin θ, which results in the equation:
This can be rearranged to produce the equation below. This equation may be used to determine the mass of a scattering atom from the kinetic energy of its ISS peak, once the primary beam energy has been calculated.
Examples
For this example, the primary ion energy is unknown. Using an instrument to acquire the scattering spectra, He+ ions of ~1 keV energy are scattered from a gold substrate. A strong peak attributable to scattering from gold is observed at 877 eV kinetic energy. Therefore, M1 = 4, M2 = 197, θ = 130°, ES = 877. The primary beam energy E0 is calculated using the first part of the equation as follows:
= 937
Thus, the primary beam energy is determined to be 937 eV.
The masses of peaks at other scattered energies can then be determined. A weak feature at 762 eV would be considered using the equation as follows:
E = ES/E0 = 762 / 937 = 0.813
C = cos 130° = -0.643
S = sin 130° = 0.766
Substituting these values results in a value for M2 of 63.7, and, therefore, the peak is from copper. The ES/E0 ratio varies with atomic weight of the surface atom and the nature of the primary ion.
Ion scattering spectra
Example ion scattering spectra from Cu, Ag and Au, using He+ ions of 970 eV energy and a scattering angle of 130°. All specimens were mildly etched to remove most of the carbon and oxygen contaminants. The copper and silver ISS spectra show some residual oxygen presence at ~400 eV KE. The Ag spectrum has some structure at ~60 eV. The low KE region of ISS corresponds to sputtered ions of Ag, which contribute to a high intensity, low kinetic energy spectrum from 0 to 200 eV KE.
whatisxps_ion-08
Primary ions in ion scattering spectroscopy
Ion scattering spectrum from a phosphor-bronze sample showing the presence of oxygen, copper and tin. This spectrum was acquired using He ions at 1 keV.
Noble gases are usually used for the primary ions in ISS, which avoids contamination of the surface that would occur if a more reactive material were used. From the above equations, the only surface atoms that can be detected are those having a greater mass than the primary ions. Helium, therefore, provides the widest mass range, and hydrogen is the only element that cannot be detected.
Because the separation of mass decreases with atomic weight, in the figure above, there is an energy separation of approximately 400 eV between O and Cu but less than 100 eV between Cu and Sn. This result is produced even though the mass difference between Sn and Cu is much greater than that between Cu and O. If better mass resolution is required, then it is necessary to use a noble gas ion that is as heavy as possible while still being lighter than the target atoms. Using argon ions, for example, on the same sample as that in the figure above can provide sufficient mass resolution to distinguish between the two copper isotopes.
Ion scattering spectrum from a phosphor-bronze sample showing the presence of oxygen, copper and tin. This spectrum was acquired using He ions at 1 keV.
Ion scattering spectrum from a phosphor-bronze sample showing the presence of oxygen, copper and tin. This spectrum was acquired using He ions at 1 keV.
Ion scattering spectroscopy mass resolution
Helium as an ion species for ISS enables the determination of elemental composition; however, unit mass resolution is lost for all target atoms heavier than Na, caused by the physics of the scattering process. Inherent FWHM of peaks near E0 are usually 2% of E. In general, mass resolution decreases rapidly with increasing mass. Mass resolution also depends on the range of angles from which ions are collected. Although the first phosphor-bronze spectrum was obtained using a collection angle of 20°, this had to be reduced to approximately 3° to obtain the second spectrum. The energy spread in the ion beam will also affect mass resolution as will the degree of collimation of the beam.
whatisxps_ion-10
Ion scattering spectrum from a phosphor-bronze sample showing the presence of two copper isotopes. This spectrum was acquired using Ar ions at 1 keV.
Ion scattering spectroscopy instrumentation
Folha de estilo para cartões de instrumentos original
Nexsa XPS
  • Módulo de inclinação para medições ARXPS
  • Fonte de íons de modo duplo para recursos de perfil de profundidade expandida
  • Análise de isolantes
K-Alpha XPS
  • Espectroscopia de área selecionável
  • Monocromador microfocalizado
  • Espectroscopia de estado químico de alta resolução
ESCALAB Xi+ XPS
  • Espectroscopia de alta sensibilidade
  • XPS com raios X não monocromáticos
  • Analisador de energia hemisférica de 180 graus
Style Sheet to change H2 style to p with em-h2-header class
Contact us