Production Bottles and Accessories
Versatile, clean, single-use bottles
Designed specifically for the storage and transport of active pharmaceutical ingredients and bulk intermediates, Thermo Scientific Nalgene production bottles and carboys offer leak-proof storage for sensitive liquids, buffers, media, and more. They are available in PC, PETG, HDPE. FEP and PFA, and in volumes ranging from 30 mL to 20 L. Extensive regulatory documentation assures that the products meet your demanding biotechnology requirements and reduces the time and cost to implement them in your process.
Key advantages of the Nalgene production bottles
Our high-quality plastic Nalgene bottles are suitable for pharmaceutical and biotechnology applications that require processing and storage of bulk intermediates and critical reagents such as vaccine and protein therapeutic preparations. The materials are biologically tested and guaranteed leakproof. Select from a variety of resin materials and bottle shapes and sizes, ranging from 0.5 mL to 50 L.
Find your production bottle for your specific application by resin type
Biotainer PETG
Eliminate the cost of cleaning, sterilizing and associated validations. Suitable for pharmaceutical and biotechnology applications that require processing and storing bulk intermediates and critical reagents such as vaccine and protein therapeutic preparations.
Ideal for prolonged storage of pH sensitive materials such as culture media. Certified sterile and ready-to-use, all Biotainer bottles are gamma irradiated to a Sterility Assurance Level (SAL) of 10–6.
Ideal for storage and sampling of active pharmaceutical ingredients and bulk intermediates PETG’s properties feature low permeability to CO2/O2 and may provide a longer-term storage solution of pH-sensitive contents.
PC
Specifically designed for pharmaceutical and biological manufacturing and to reduce the risk of carry-over contamination. Durable, translucent PC construction ensures the integrity of your sample, even frozen to -100 °C
HDPE
Our new certified platinum clean products include particulate certifications to under 1/3 the allowable limits according to USP <788> and lower endotoxin levels of 0.25 EU/mL, to help mitigate potential sources of unwanted impurities.
These bottles ideal for storage and transport of biological materials and offer excellent chemical resistance and support storage conditions of -100° to +120°C. Biotainers are certified sterile and ready to use.
LSP_VaB_FEP-Teflon
Excellent for storing high purity chemicals, this product includes a certificate of quality that assures the product has been tested and accepted in accordance with specifications.
Perform trace metal analysis and organic solvent applications with temperature and chemical-resistant Thermo Scientific Nalgene Narrow-Mouth FEP Bottles.
Perform trace metal analysis and organic solvent applications with the Nalgene Wide-Mouth temperature and chemical-resistant FEP bottles.
FEP
Use at high and low temperatures for trace metal analysis and applications with organic solvents. Specially designed for US EPA Method 1311: Toxicity Characteristic Launching Program (TCLP), a safer alternative without sacrificing needed accuracy.
T_140218_224
Get excellent resistance to temperature, chemicals and corrosion with the narrow-mouth bottles that among the most chemical- and corrosion-resistant containers available
Biotainer&#x20;PETG
Eliminate the cost of cleaning, sterilizing and associated validations. Suitable for pharmaceutical and biotechnology applications that require processing and storing bulk intermediates and critical reagents such as vaccine and protein therapeutic preparations.
Ideal for prolonged storage of pH sensitive materials such as culture media. Certified sterile and ready-to-use, all Biotainer bottles are gamma irradiated to a Sterility Assurance Level (SAL) of 10–6.
Ideal for storage and sampling of active pharmaceutical ingredients and bulk intermediates PETG’s properties feature low permeability to CO2/O2 and may provide a longer-term storage solution of pH-sensitive contents.
PC
Specifically designed for pharmaceutical and biological manufacturing and to reduce the risk of carry-over contamination. Durable, translucent PC construction ensures the integrity of your sample, even frozen to -100 °C
HDPE
Our new certified platinum clean products include particulate certifications to under 1/3 the allowable limits according to USP <788> and lower endotoxin levels of 0.25 EU/mL, to help mitigate potential sources of unwanted impurities.
These bottles ideal for storage and transport of biological materials and offer excellent chemical resistance and support storage conditions of -100° to +120°C. Biotainers are certified sterile and ready to use.
LSP_VaB_FEP-Teflon
Excellent for storing high purity chemicals, this product includes a certificate of quality that assures the product has been tested and accepted in accordance with specifications.
Perform trace metal analysis and organic solvent applications with temperature and chemical-resistant Thermo Scientific Nalgene Narrow-Mouth FEP Bottles.
Perform trace metal analysis and organic solvent applications with the Nalgene Wide-Mouth temperature and chemical-resistant FEP bottles.
FEP
Use at high and low temperatures for trace metal analysis and applications with organic solvents. Specially designed for US EPA Method 1311: Toxicity Characteristic Launching Program (TCLP), a safer alternative without sacrificing needed accuracy.
T_140218_224
Get excellent resistance to temperature, chemicals and corrosion with the narrow-mouth bottles that among the most chemical- and corrosion-resistant containers available
Related products
Ordering information
PETG
PC
HDPE
FEP
Accessories
Bioprocessing resources
Click below to access products brochures, training videos, mobile apps, and more.