    • See all product categories
  • Applications
    • Bioprocessing
  • Services
    • 360° CDMO and CRO Solutions
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • FBXO2 Antibodies
          Invitrogen
          FBXO2 Polyclonal Antibody
          View all (8) FBXO2 antibodies
          Questions & Answers
          Cite FBXO2 Polyclonal Antibody
          Additional Information:
          {{banner.description}}
          • Antibody Testing Data (2)
          FBXO2 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          FBXO2 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          FIGURE: 1 / 2
          FBXO2 Antibody (PA5-23718) in WB
          Western blot analysis of FBXO2 in mouse brain tissue and HepG2, 293 cell line lysates. Samples were incubated with FBXO2 polyclonal antibody (Product # PA5-23718). Lysates: 35 µg/lane. FBXO2 protein (arrow). {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product
          {{$ctrl.videoDesc}}
          {{$ctrl.videoLongDesc}}
          FBXO2 Antibody in Western Blot (WB)
          FBXO2 Antibody in Flow Cytometry (Flow)
          Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.
          FBXO2 Polyclonal Antibody
          Product Details
          PA5-23718
          Applications
          Tested Dilution
          Publications
          Western Blot (WB)
          1:1,000
          -
          Flow Cytometry (Flow)
          1:10-1:50
          -
          Product Specifications
          Species Reactivity
          Human, Mouse
          Host/Isotype
          Rabbit / IgG
          Class
          Polyclonal
          Type
          Antibody
          Immunogen
          KLH conjugated synthetic peptide between 244-271 amino acids from the C-terminal region of human FBXO2
          3D Epitope / Immunogen
          Conjugate
          Unconjugated Unconjugated Unconjugated
          Form
          Liquid
          Concentration
          0.5 mg/mL
          Amount
          200 µg
          Purification
          Protein A, Antigen affinity chromatography
          Storage buffer
          PBS, pH 7.4
          Contains
          0.09% sodium azide
          Storage conditions
          Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
          Shipping conditions
          Ambient (domestic); Wet ice (international)
          RRID
          AB_2541218
          Target Information
          This gene encodes a member of the F-box protein family which is characterized by an approximately 40 amino acid motif, the F-box. The F-box proteins constitute one of the four subunits of the ubiquitin protein ligase complex called SCFs (SKP1-cullin-F-box), which function in phosphorylation-dependent ubiquitination. The F-box proteins are divided into 3 classes: Fbws containing WD-40 domains, Fbls containing leucine-rich repeats, and Fbxs containing either different protein-protein interaction modules or no recognizable motifs. The protein encoded by this gene belongs to the Fbxs class. This protein is highly similar to the rat NFB42 (neural F Box 42 kDa) protein which is enriched in the nervous system and may play a role in maintaining neurons in a postmitotic state.
          Bioinformatics
          Protein Aliases: F-box only protein 2; prion protein ligand 4
          View more View less
          Gene Aliases: FBG1; Fbs1; Fbs2; FBX2; Fbxo2; NFB42; Prpl4
          View more View less
          UniProt ID: (Mouse) Q80UW2
          View more View less
          Entrez Gene ID: (Mouse) 230904
          View more View less
          Function(s)
          beta-amyloid binding glycoprotein binding protein binding carbohydrate binding
          Process(es)
          ubiquitin-dependent protein catabolic process glycoprotein catabolic process negative regulation of cell proliferation protein ubiquitination ER-associated ubiquitin-dependent protein catabolic process SCF-dependent proteasomal ubiquitin-dependent protein catabolic process regulation of protein ubiquitination
          Your items have has been added!
          Host server : magellan-search-green-dd86746ff-t8vjm:80/100.66.78.50:80.
          git-commit: 04e44c58f20fafe66519586ffe6db5d8a90e52ce
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.53.1-Offline