    • See all product categories
  • Applications
    • Bioprocessing
  • Services
    • 360° CDMO and CRO Solutions
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • MHC Class II Antibodies
          Fabgennix
          MHC Class II Polyclonal Antibody
          View all (19) MHC Class II antibodies
          Questions & Answers
          Cite MHC Class II Polyclonal Antibody
          Additional Information:
          {{banner.description}}
          Product Details
          MHCII-101AP
          Applications
          Tested Dilution
          Publications
          Western Blot (WB)
          1:500
          -
          ELISA (ELISA)
          1:50,000
          -
          Immunoprecipitation (IP)
          1:200
          -
          Product Specifications
          Species Reactivity
          Human, Mouse, Rat
          Host/Isotype
          Rabbit / IgG
          Class
          Polyclonal
          Type
          Antibody
          Immunogen
          Synthetic peptide taken within amino acid region 1-50 on mouse Major Histocomatibility Complex Class II antigen (MHC II).
          Conjugate
          Unconjugated Unconjugated Unconjugated
          Form
          Liquid
          Concentration
          0.5-1.5 mg/mL
          Purification
          Affinity chromatography
          Storage buffer
          proprietary buffer, pH 7.4-7.8, with 0.5% BSA, 30% glycerol
          Contains
          0.02% sodium azide
          Storage conditions
          -20°C
          Shipping conditions
          Ambient (domestic); Wet ice (international)
          Target Information
          MHC (major histocompatibility complex) class II molecules are transmembrane glycoproteins expressed on the surface of professional antigen-presenting cells, such as macrophages, dendritic cells and B cells. Before their exposition on the cell surface, the MHC class II molecules react with endocytosed exogenous antigens, which are then presented to the T cells. The antigen-binding grove between MHC class II alpha and beta chain is open at both ends and is 15-24 amino acid residues long.
          HLA and MHC antibodies play a significant role in Immunopeptidomics, facilitating the identification and characterization of neoantigens through high-performance liquid chromatography coupled to tandem Mass Spectrometry.
          Bioinformatics
          Protein Aliases: MHC 2; MHC Class II; MHC II; MHCCII
          Your items have has been added!
          Host server : magellan-search-green-dd86746ff-hszvs:80/100.66.78.131:80.
          git-commit: 04e44c58f20fafe66519586ffe6db5d8a90e52ce
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.53.1-Offline