Search
Copeptin is a reliable surrogate marker of vasopressin (AVP/ADH), helping overcome the technical difficulties and limited reliability of available vasopressin assays. Copeptin can be utilized alongside clinical evaluation and other laboratory findings to provide a multitude of benefits to patients, including serving as a diagnostic aid when combined with either conventional or high-sensitivity Troponin for ruling out acute myocardial infarction (AMI) and differentially diagnosing polyuria-polydipsia syndrome.
Key benefits of the Thermo Scientific B·R·A·H·M·S Copeptin ProAVP KRYPTOR Assay include:
Thermo Fisher Scientific products are distributed globally and their uses, applications, indications, claims and availability of products in each country depend on local regulatory marketing authorization status, please consult the Instructions For Use (IFU) available in your country.
© 2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified. B·R·A·H·M·S and B·R·A·H·M·S Copeptin proAVP are registered trademarks of B·R·A·H·M·S GmbH.
KRYPTOR and TRACE are trademarks of Cisbio Bioassays, licensed for use by B·R·A·H·M·S GmbH, a part of Thermo Fisher Scientific. Nobel Prize is a registered trademark of the Nobel Foundation. Other product names in this document are used for identification purposes; they may be trademarks and/or registered trademarks of their respective companies.