Search
In the rapidly evolving field of precision oncology, the increasing availability of targeted treatments offers renewed hope to patients. Unfortunately, the lengthy wait for next-generation sequencing (NGS) results—often weeks—and the risk of insufficient sample quantity (QNS) can diminish this hope. The newly FDA-approved Oncomine Dx Express Test is set to revolutionize this process. Capable of being implemented in labs without NGS expertise, and close to patients, it delivers results within just 24 hours—from tissue sample to report.
Deliver NGS results in as little as 24 hours from tissue sample to report
The Oncomine Dx Express Test can eliminate the lengthy wait for a complete biomarker picture. It enables pathology labs to deliver results in as little as 24 hours, facilitating faster, more informed treatment decisions so that your patients don't have to wait weeks for results.
Bring precision oncology close to your patients
The Oncomine Dx Express Test is designed for implementation in local laboratories, enabling in-house testing. This supports timely, decentralized precision oncology by delivering NGS results close to patients.
Get results for more patients, even with a small sample size
The Oncomine Dx Express Test requires only 10 ng of DNA and RNA, which is significantly less sample input than other NGS technologies and therefore is more efficient even with challenging samples and small biopsies.
Angie Cheng
Director, Clinical R&D
Thermo Fisher Scientific
Abstract:
Angie Cheng presented the analytical and clinical validation of the Oncomine™ Dx Express Test, a targeted next-generation sequencing (NGS) in vitro diagnostic assay designed for companion diagnostic (CDx) and comprehensive tumor profiling applications. The data demonstrate strong performance across limit of blank (LoB), limit of detection (LoD), reproducibility, repeatability, and analytical accuracy studies, with high agreement (>90–97%) compared to orthogonal methods and robust detection across variant types. Clinical results further support its use as a companion diagnostic for EGFR exon 20 insertions in NSCLC to guide targeted therapy selection.
David Chi
Director of Oncology Product
Management, Thermo Fisher Scientific
Abstract:
David Chi presented the newly FDA-approved Oncomine™ Dx Express Test (ODxET)—a comprehensive next-generation sequencing (NGS) solution for solid tumor profiling and companion diagnostics. The presentation highlighted the test’s ability to analyze 46 genes across DNA and RNA, including EGFR exon 20 insertion as a CDx indication. Built on the Genexus™ Dx Integrated Sequencer, the test delivers results in as little as 24 hours with only ~20 minutes of hands-on time, significantly simplifying NGS implementation for community and hospital laboratories. With automated workflow, flexible multiplexing, and payer-aligned validation, the ODxET addresses key barriers to NGS adoption—turnaround time, cost, complexity, and staffing—enabling rapid, reliable biomarker-driven treatment decisions.
Gary Pestano, PhD
Chief Development Officer
Biodesix, Inc.
Abstract:
Gary Pestano of Biodesix shared their real-world experience evaluating the Oncomine Dx Express Test and Genexus Dx System in a CAP/CLIA-certified laboratory setting. Using 48 FFPE samples, the team demonstrated 98% variant-level concordance compared to a validated reference NGS assay, underscoring the test’s accuracy and reproducibility. Laboratory staff praised the system’s end-to-end automation, dual-touch workflow, and ability to process up to 12 samples per run with rapid 24–48-hour turnaround. Feedback emphasized major workflow efficiencies—from automated extraction to integrated bioinformatics reporting—making ODxET a practical and scalable solution for molecular diagnostics laboratories seeking to expand NGS capabilities.
Bence Sipos, MD
Molecular Pathology
Baden-Württemberg, Stuttgart, Germany
Abstract:
Prof. Bence Sipos shared practical, pathology-centered insights into adopting rapid NGS for oncology testing. He discussed the growing clinical need for comprehensive molecular characterization from minimal FFPE samples and highlighted the Oncomine Dx Express Test as a solution balancing speed, sensitivity, and sample efficiency. His laboratory in Stuttgart performed ~5,000 NGS tests annually, achieving ≤3-day turnaround in over 80% of cases and <1% drop-off rate. Case studies in NSCLC and other solid tumors illustrated ODxET’s diagnostic value in guiding timely targeted therapy decisions. Dr. Sipos concluded that rapid, reliable NGS is now equivalent in turnaround and dependability to traditional pathology methods like IHC—representing a “quiet revolution” in molecular diagnostics.
Steve Stone
Managing Director
Argent Global Services
Abstract:
See how automation and workflow design impact speed, efficiency, and cost in real‑world NGS testing.
As oncology testing demand grows, labs are under pressure to deliver accurate results faster with fewer hands and tighter budgets. In this webinar, you’ll see real time‑and‑motion data comparing two NGS workflows (amplicon- based Oncomine Dx Express Test and hybrid capture based NGS workflow) for routine precision oncology, and learn how automation can reduce touchpoints, shorten turnaround time, and support sustainable scaling. You’ll walk away with clarity on:
Table 1. Companion diagnostic indications
Tissue type |
Gene |
Variant |
Targeted therapy |
Non-small cell lung cancer (NSCLC) |
EGFR |
EGFR exon 20 insertions |
ZEGFROVY™ (sunvozertinib) |
Table 2. Oncomine Dx Express Test panel gene list for tumor profiling
DNA |
RNA |
|||||||||
Substitutions, insertions, and deletions |
Copy number variants |
Fusions and splice variants |
||||||||
AKT1 AKT2 AKT3 ALK AR ARAF BRAF |
CDK4 CHEK2 CTNNB1 EGFR ERBB2 ERBB3 ERBB4 |
ESR1 FGFR1 FGFR2 FGFR3 FGFR4 FLT3 GNAS |
HRAS IDH1 IDH2 KEAP1 KIT KRAS MAP2K1 |
MAP2K2 MET NRAS NTRK1 NTRK2 NTRK3 PDGFRA |
PIK3CA PTEN RAF1 RET ROS1 STK11 TP53 |
AR EGFR ERBB2 ERBB3 FGFR1 |
FGFR2 FGFR3 KRAS MET PIK3CA |
ALK AR BRAF EGFR ESR1 FGFR1 |
FGFR2 FGFR3 MET NRG1 NTRK1 NTRK2 |
NTRK3 NUTM1 RET ROS1 RSPO2 RSPO3 |
The Genexus Dx System automates the NGS workflow from patient samples to reports with only two user touchpoints and as little as 20 minutes of hands-on time.
Pre-treated FFPE tissue sections are loaded onto the Genexus Dx Purification System and the instrument performs automated purification and quantification of DNA and RNA.
Library preparation, template preparation, sequencing, analysis, and reporting are performed by the Genexus Dx Integrated Sequencer. Throughout this procedure, sample and reagent information is recorded and tracked by one integrated software. This highly automated workflow helps reduce laboratory staff burden and the potential for human errors and alleviates the need for specialized bioinformatics expertise.
Oncomine Dx Express Test workflow
We will be happy to answer your questions about bringing NGS to your laboratory.
For In Vitro Diagnostic Use.
CN: 63025