Search
Find valuable information.
Optimize your experiments to get the best results. We’ve compiled a detailed knowledge base of the top tips and tricks to meet your research needs.
View the relevant questions below:
| Recommended Maintenance Schedule | |
|---|---|
| Power on/off the computer controlling the instrument | Weekly |
| Check computer disk space. If necessary, archive or back up your experiment files and instrument settings | Weekly |
| Background calibration | Every month |
| Run disk cleanup and disk defragmentation | Every month |
| Perform an instrument self test | Every month |
| Pure dye calibrations | Every 18 months |
| Spatial calibration | Every 18 months |
| RNaseP instrument verification | After the instrument has been moved, or as needed to verify instrument performance |
The RNaseP verification plate contains template, master mix, and a TaqMan® assay for RNaseP. It is used to verify that the instrument is performing to specifications. If you have reason to suspect there is something wrong with the instrument, or if you want to rule out a chemistry issue, the RNaseP plate is a good way to test the system. It is also recommended to use after the instrument has been moved to a new location. The RNaseP verification plate is a single-use plate.
The calibration plates can be stored and reused three times for up to one year after you receive them, so make sure to return them to their original packaging and return them to –20°C storage until the next use. If needed, you can make your own background plate using deionized water. Please follow the directions in the StepOne™ and StepOnePlus™ Maintenance and Administration Guide (Appendix C) for more details.
The StepOne™ and StepOnePlus™ Real-Time PCR Systems have a Fast block, which supports reaction volumes of 10–30 µL.
Follow these instructions for proper loading of tubes, tube strips, and plates.
For optimal performance on the StepOnePlus™ instrument with partial loads:
For optimal performance on the StepOne™ instrument, load at least 4 tubes in the sample block.
Please refer to this selection table for compatible plates, tubes, and films.
The StepOnePlus™ Real-Time PCR System contains six independently thermally regulated VeriFlex™ blocks, which can help you optimize your thermal cycling conditions. You can set a different temperature for one or more zones, or you can set one temperature for all zones in the sample blocks. [Note: The difference in temperature between adjacent zones can range from 0 to 5°C, in 0.1°C increments].
In a touchdown PCR experiment, you will change either the temperature or the time of a particular PCR step with every cycle. Most commonly, the annealing temperature is adjusted throughout the experiment, such that the specificity is higher in the early cycles and the efficiency in the later cycles. In this example, we will set the method to do the following:
40 cycles ![]() |
|
Run files will be saved to both the instrument and the connected computer. On the connected computer, files will be saved to the default data folder, unless you change it.
To find or change the default folder, go to Tools → Preferences → Defaults. Here you will see a Data Folder and an Import Folder. The default location is shown. If you want files to be saved to (or open from) a different location, click ‘Browse’ and choose the new folder.
For genotyping data, we recommend TaqMan® Genotyper Software. For relative quantitation, we recommend ExpressionSuite™ Software.
For Research Use Only. Not for use in diagnostic procedures.