    • See all product categories
  • Applications
    • Bioprocessing
  • Services
    • 360° CDMO and CRO Solutions
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • ELISA Kits ›
          • AKT1 ELISA Kits
          Invitrogen
          Multispecies AKT (Phospho) [pS473] InstantOne™ ELISA Kit
          1 Reference
          View all (4) AKT1 ELISA Kits
          Technical guide & protocol Tech Support
          Technical guide & protocol Tech Support
          Technical guide & protocol Tech Support
          Technical guide & protocol Tech Support
          Additional Information:
          {{banner.description}}
          Multispecies AKT (Phospho) [pS473] InstantOne™ ELISA Kit
          Group 53 Created with Sketch.
          Multispecies AKT (Phospho) [pS473] InstantOne™ ELISA Kit
          Group 53 Created with Sketch.
          FIGURE: 1 / 3
          Multispecies AKT (Phospho) [pS473] InstantOne™ ELISA Kit
          Representative data for AKT (Phospho) [pS473] Multispecies InstantOne™ ELISA. MCF7 cells were pre-treated with 10 µM wortmannin washed and unstimulated (-) or stimulated with 10 ng/mL insulin for 10 min (+). Cells were analyzed by immunoblotting (left) and InstantOne™ ELISA (right). The intensity of the band at 56 kDa as detected by immunoblot correlates with InstantOne™ ELISA absorption values. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product
          {{$ctrl.videoDesc}}
          {{$ctrl.videoLongDesc}}
          Multispecies AKT (Phospho) [pS473] InstantOne™ ELISA Kit
          Multispecies AKT (Phospho) [pS473] InstantOne™ ELISA Kit (85-86042-253)
          videoThumbnail
          ►
          Product Specifications
          Sample type/volume
          Cell Lysate
          20 µL
          Hands-on time
          1 hr 20 min
          Time-to-result
          1 hr 10 min
          Homogenous (no wash)
          No
          Instrument
          Absorbance Microplate Reader
          Product size
          25 x 384 Tests
          Contents
          Capture Antibody
          Detection Antibody Reagent
          InstantOne™ Assay Plate
          Cell Lysis Buffer
          Enhancer Solution
          Wash Buffer (10X)
          Substrate Reagent (TMB)
          Stop Solution
          Positive Control Cell Lysate (lyophilized)
          Plate seal
          Shipping conditions
          Wet ice
          Storage
          2-8°C
          Protein name
          AKT1
          Target site-specificity
          Phosphorylated Ser473
          Protein subtype
          Adaptor Proteins
          Species (tested)
          Human
          Assay kit format
          InstantOne ELISA
          Detector antibody conjugate
          Biotin
          Label or dye
          HRP
          About This Kit
          The InstantOne™ ELISA is specifically engineered for accurate measurement of phosphorylated human, mouse, and hamster AKT 1/2/3 in cell lysates. Detection of rat AKT 1/2/3 is expected. The InstantOne™ ELISA kit allows for fast analysis of samples in approximately one hour. All reagents used in a traditional sandwich ELISA are added in solution to a plate followed by a wash step and detection with the TMB colorimetric substrate.
          Target information
          AKT1 (PKB alpha) is a serine/threonine kinase that regulates cell survival. The activated enzyme inhibits apoptosis and stimulates cell cycle progression by phosphorylating numerous targets in various cell types, including cancer cells. This protein kinase is activated by insulin, PI3K, IGF1 and various other growth and survival factors. Akt promotes cell survival by inhibiting apoptosis through phosphorylation and inactivation of several targets, including forkhead transcription factors, and caspase-9. The AKT pathway is a major target for cancer drug discovery.
          Bioinformatics
          Gene aliases : AKT, AKT1, PKB, PKB-ALPHA, PRKBA, RAC, RAC-ALPHA
          Gene ID : (Human) 207
          Gene symbol : AKT1
          Protein Aliases : AKT, AKT1M, CAKT, C-AKT, PKB, PKB ALPHA, RAC-PK-gamma, V-AKT MURINE THYMOMA VIRAL ONCOGENE HOMOLOG 1, V-AKT MURINE THYMOMA VIRAL ONCOGENE-LIKE PROTEIN 1, PROTO-ONCOGENE C-AKT, RAC, RAC-ALPHA, RAC-ALPHA SERINE/THREONINE-PROTEIN KINASE, RAC-PK-ALPHA, RAC-PK-beta, PKB beta, PKB gamma, PKB-ALPHA, PKBG, PRKBA, PROTEIN KINASE B ALPHA, AKT1m, PKB alpha, Protein kinase B, Protein kinase B alpha, Proto-oncogene c-Akt, rac protein kinase alpha, RAC-alpha serine/threonine-protein kinase, RAC-PK-alpha, serine-threonine protein kinase, unnamed protein product, v-akt murine thymoma viral oncogene homolog 1, v-akt murine thymoma viral oncogene-like protein 1
          UniProt ID (Human) P31749
          Recommended Products
          IL-6 Human Matched Antibody Pair
          Because you are interested in IL-6 Human ELISA Kit.
          SIZE
          10 x 96 tests
          PRICE
          USD 400.00
          Cat # CHC1263
          guarantee_icon
          Performance Guarantee
          If an Invitrogen™ immunoassay doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          Your items have has been added!
          Host server : magellan-search-green-6b469b8bb7-kg5vd:80/100.66.78.64:80.
          git-commit: a334af76dff23450325448aedefe62379591458a
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.48.1-2026.04.16-1.0