    • See all product categories
  • Applications
    • Bioprocessing
  • Services
    • 360° CDMO and CRO Solutions
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • ELISA Kits ›
          • TRAIL (CD253) ELISA Kits
          This product was previously sold as SKU 900-K141
          Invitrogen
          Human TRAIL (Soluble) ELISA Development Kit (ABTS), PeproTech®
          2 References
          View all (3) TRAIL (CD253) ELISA Kits
          Technical guide & protocol Tech Support
          Technical guide & protocol Tech Support
          Technical guide & protocol Tech Support
          Technical guide & protocol Tech Support
          {{$ctrl.translations['antibody.pdp.skuNotes.additionalInformation']}}:
          {{$ctrl.translations[$ctrl.isSkuNotesCollapsed ? 'antibody.pdp.skuNotes.viewMore' : 'antibody.pdp.skuNotes.viewLess']}}
          Human TRAIL (Soluble) ELISA Development Kit (ABTS)
          Group 53 Created with Sketch.
          Human TRAIL (Soluble) ELISA Development Kit (ABTS)
          Group 53 Created with Sketch.
          FIGURE: 1 / 3
          Human TRAIL (Soluble) ELISA Development Kit (ABTS)
          Human TRAIL (CD253) ABTS ELISA standard curve (Product # 900-K141K). {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product
          {{$ctrl.videoDesc}}
          {{$ctrl.videoLongDesc}}
          Human TRAIL (Soluble) ELISA Development Kit (ABTS)
          Human TRAIL (Soluble) ELISA Development Kit (ABTS) (900-K141K)
          videoThumbnail
          ►
          Product Specifications
          Analytical sensitivity
          Lot-specific
          Assay range
          Lot-specific
          Hands-on time
          1 hr 45 min
          Time-to-result
          24 hr 30 min
          Homogenous (no wash)
          No
          Instrument
          Absorbance Microplate Reader
          Product size
          10 x 96 Tests
          Contents
          This Human TRAIL (CD253) ELISA Kit includes Capture Antibody, Detection Antibody, Protein Standard, and HRP-conjugate. The HRP-conjugate is packaged separately.
          Storage
          Refer to product documentation for component specific storage temperature.
          Protein name
          TRAIL (CD253)
          Protein family
          Growth Factors & Receptors
          Species (tested)
          Human
          Detector antibody conjugate
          Biotin
          Label or dye
          HRP
          About This Kit
          900-K141K consists of: 900-K141 (capture antibody, detection antibody, and protein standard), and AVIDIN-60UL (avidin HRP-conjugate).
          Human TRAIL (CD253) ABTS ELISA Development Kit contains the key components required for the quantitative measurement of natural and/or recombinant human TRAIL (CD253) in a sandwich ELISA format. Human TRAIL (CD253) ABTS ELISA Development Kit contains enough material to assay the target cytokine in at least 1,000 ELISA plate wells.
          The ABTS ELISA Buffer Kit (Product # 900-K00), sold separately, is recommended to be used in conjunction with this ABTS ELISA Development Kit for optimal performance. This buffer kit contains all the necessary components to assay ten 96-well ELISA plates (included) and contains detailed handling instructions.
          Target information
          TRAIL is a cytokine that belongs to the tumor necrosis factor (TNF) ligand family. TRAIL preferentially induces apoptosis in transformed and tumor cells, but does not appear to kill normal cells although it is expressed at a significant level in most normal tissues. TRAIL binds to several members of TNF receptor superfamily including TNFRSF10A/TRAILR1, TNFRSF10B/TRAILR2, TNFRSF10C/TRAILR3, TNFRSF10D/TRAILR4, and possibly also to TNFRSF11B/OPG. The activity of this protein may be modulated by binding to the decoy receptors TNFRSF10C/TRAILR3, TNFRSF10D/TRAILR4, and TNFRSF11B/OPG that cannot induce apoptosis. The binding of TRAIL protein to its receptors has been shown to trigger the activation of MAPK8/JNK, caspase 8, and caspase 3. Receptors for TRAIL include two death domain containing receptors, death receptor 4 (DR4) and DR5, as well as two decoy receptors, DcR1 and DcR2, lacking the intracellular signaling death domain. TRAIL is a type II membrane protein and expressed in a variety of human tissues. Like TNF and Fas ligand, TRAIL induces apoptosis and NF-kB activation in many tissues and cells.
          Bioinformatics
          Gene aliases : Apo-2L, APO2L, CD253, TANCR, TL2, TNLG6A, TRAIL
          Gene ID : (Human) 8743
          Gene symbol : TNFSF10
          Protein Aliases : Apo-2 ligand, CD253 antigen, TN, TNF superfamily member 10, TRAIL protein, chemokine tumor necrosis factor ligand superfamily member 10, TNF related apoptosis inducing ligand (TRAIL)-short, TNF-related apoptosis inducing ligand TRAIL, TNFSF10, TRAIL splice variant delta, tumor necrosis factor (ligand) family, member 10, tumor necrosis factor (ligand) superfamily, tumor necrosis factor ligand 6A, tumor necrosis factor superfamily member 10, unnamed protein product
          UniProt ID (Human) P50591
          Recommended Products
          IL-6 Human Matched Antibody Pair
          Because you are interested in IL-6 Human ELISA Kit.
          SIZE
          10 x 96 tests
          PRICE
          USD 400.00
          Cat # CHC1263
          guarantee_icon
          Performance Guarantee
          If an Invitrogen™ immunoassay doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          Your items have has been added!
          Host server : magellan-search-blue-6b866bbc8c-mwps6:80/100.66.76.64:80.
          git-commit: 9d76681b284ad04e8ddcd97b79462898b32e8e5b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.61.1-2026.10.99-1.0