    • See all product categories
  • Applications
    • Cell Analysis
    • Protein Biology
    • Lab Informatics
    • 360° CDMO and CRO Solutions
  • Help and Support
    • Customer Center
    • Contact Us
    • Certificates of Analysis and Conformance
    • Safety Data Sheets (SDS)
    • Manuals
    • How to Cite Our Products in a Paper
    • Instrument Support
    • Knowledge Base and Product FAQs
            • Services Central
          • ELISA Kits ›
          • BMP-2 ELISA Kits
          This product was previously sold as SKU 900-K255
          Invitrogen
          Human/Mouse/Rat BMP-2 ELISA Development Kit (ABTS), PeproTech®
          12 References
          View all (3) BMP-2 ELISA Kits
          Technical guide & protocol Tech Support
          Technical guide & protocol Tech Support
          Technical guide & protocol Tech Support
          Technical guide & protocol Tech Support
          {{$ctrl.translations['antibody.pdp.skuNotes.additionalInformation']}}:
          {{$ctrl.translations[$ctrl.isSkuNotesCollapsed ? 'antibody.pdp.skuNotes.viewMore' : 'antibody.pdp.skuNotes.viewLess']}}
          Human/Mouse/Rat BMP-2 ELISA Development Kit (ABTS)
          Group 53 Created with Sketch.
          Human/Mouse/Rat BMP-2 ELISA Development Kit (ABTS)
          Group 53 Created with Sketch.
          FIGURE: 1 / 3
          Human/Mouse/Rat BMP-2 ELISA Development Kit (ABTS)
          Human Mouse Rat BMP-2 ABTS ELISA standard curve (Product # 900-K255K). {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product
          {{$ctrl.videoDesc}}
          {{$ctrl.videoLongDesc}}
          Human/Mouse/Rat BMP-2 ELISA Development Kit (ABTS)
          Human/Mouse/Rat BMP-2 ELISA Development Kit (ABTS) (900-K255K)
          videoThumbnail
          ►
          Product Specifications
          Analytical sensitivity
          Lot-specific
          Assay range
          Lot-specific
          Hands-on time
          1 hr 45 min
          Time-to-result
          24 hr 30 min
          Homogenous (no wash)
          No
          Instrument
          Absorbance Microplate Reader
          Product size
          10 x 96 Tests
          Contents
          This Human/Mouse/Rat BMP-2 ELISA Kit includes Capture Antibody, Detection Antibody, Protein Standard, and HRP-conjugate. The HRP-conjugate is packaged separately.
          Storage
          Refer to product documentation for component specific storage temperature.
          Protein name
          BMP-2
          Protein family
          Growth Factors & Receptors
          Species (tested)
          Human, Mouse, Rat
          Detector antibody conjugate
          Biotin
          Label or dye
          HRP
          About This Kit
          900-K255K consists of: 900-K255 (capture antibody, detection antibody, and protein standard), and AVIDIN-60UL (avidin HRP-conjugate).
          Human/Mouse/Rat BMP-2 ABTS ELISA Development Kit contains the key components required for the quantitative measurement of natural and/or recombinant human/mouse/rat BMP-2 in a sandwich ELISA format. Human/Mouse/Rat BMP-2 ABTS ELISA Development Kit contains enough material to assay the target cytokine in at least 1,000 ELISA plate wells.
          The ABTS ELISA Buffer Kit (Product # 900-K00), sold separately, is recommended to be used in conjunction with this ABTS ELISA Development Kit for optimal performance. This buffer kit contains all the necessary components to assay ten 96-well ELISA plates (included) and contains detailed handling instructions.
          Target information
          Bone Morphogenic Proteins (BMP) are members of the TGF-beta superfamily that affect bone and cartilage formation (Hogan 1996, Reddi 1998 and Francis-West et al. 1999). Mature BMPs are 30-38 kDa proteins that assume a TGF-beta -like cysteine knot configuration. Lovostatin increases bone formation by turning on the bmp-2 gene (Mundy et al. 1999). BMPs stimulate the production of specific bone matrix proteins and alter stromal cell and osteoclast proliferation (Macias et al. 1999, Lecanda et al. 1997). BMPs may also be an important factor for development of the viscera, with roles in cell proliferation, apoptosis, differentiation, and morphogenesis (Hogan 1996, Dale and Wardle 1999). BMPs appear to be responsible for normal dorsal/ventral patterning. Like TGF-beta, BMPs bind to a type II receptor, which then recruits the transducing type I receptor unit, activating the Smad protein signaling pathway (Massague 1994, Derynck 1997, Attisano 1993).
          Bioinformatics
          Gene aliases : BDA2, BMP2A, SSFSC, SSFSC1
          Gene ID : (Human) 650, (Mouse) 12156, (Rat) 29373
          Gene symbol : BMP2, Bmp2, Bmp2
          Protein Aliases : 2610024H22Rik, AL117858, AW546137, BB189135, BMP, BMPRII, BMPR-II, Bone morphogenetic, Bone morphogenetic protein, H-BMP-2, BMP-2, BMP-2A, bone morphogenetic protein 2A, osteogenic protein 2, unnamed protein product
          UniProt ID (Human) P12643, (Mouse) P21274, (Rat) P49001
          Recommended Products
          IL-6 Human Matched Antibody Pair
          Because you are interested in IL-6 Human ELISA Kit.
          SIZE
          10 x 96 tests
          PRICE
          USD 400.00
          Cat # CHC1263
          guarantee_icon
          Performance Guarantee
          If an Invitrogen™ immunoassay doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
          • Trademarks
          • Fair Trade Fair Trade
          Korea flag icon
          Korea
          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담
          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat
          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담
          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat
          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담
          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat
          Thermo Fisher Scientific Korea Ltd.
          Representative : Soojin Seok
          Company Registration No. : 117-81-46910
          Thermo Fisher Scientific Solutions LLC
          Representative : Soojin Seok
          Company Registration No. : 114-86-04783
          Location: 12F Suseo Office Building, 281 Gwangpyeong-ro, Gangnam-gu, Seoul, Korea(06349) | Mail-Order Business Registration : 2015-Seoul Gangnam-00898 | Payment : ShinHan Bank 140-004-396660 (Thermo Fisher Scientific Solutions LLC)
          ISMS Logo
          Your items have has been added!
          Host server : magellan-search-blue-6b866bbc8c-d6wk2:80/100.66.72.150:80.
          git-commit: 9d76681b284ad04e8ddcd97b79462898b32e8e5b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.61.1-2026.10.99-1.0