    • See all product categories
  • Applications
    • Bioprocessing
  • Services
    • 360° CDMO and CRO Solutions
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • ELISA Kits ›
          • CCL8 (MCP2) ELISA Kits
          Invitrogen
          Human MCP-2 ProQuantum Immunoassay Kit
          View all (3) CCL8 (MCP2) ELISA Kits
          Technical guide & protocol Tech Support
          Technical guide & protocol Tech Support
          Technical guide & protocol Tech Support
          Technical guide & protocol Tech Support
          {{$ctrl.translations['antibody.pdp.skuNotes.additionalInformation']}}:
          {{$ctrl.translations[$ctrl.isSkuNotesCollapsed ? 'antibody.pdp.skuNotes.viewMore' : 'antibody.pdp.skuNotes.viewLess']}}
          Human MCP-2 ProQuantum Immunoassay Kit
          Group 53 Created with Sketch.
          Human MCP-2 ProQuantum Immunoassay Kit
          Group 53 Created with Sketch.
          FIGURE: 1 / 4
          Human MCP-2 ProQuantum Immunoassay Kit
          How ProQuantum immunoasssays work
          (A) Antibody-antigen binding: Antibodies bind to two separate epitopes on the antigen (during a 1-hr incubation), which brings the two conjugated oligonucleotides into close proximity. (B) Ligation and amplification of signal (in a qPCR instrument): DNA ligase and a third splint oligonucleotide connector are added to ligate the two ends of the conjugated oligonucleotides, creating a 100-base DNA template. Once the ligase is inactivated at 95°C, the sample is amplified through 40 cycles of annea... View More
          {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product
          {{$ctrl.videoDesc}}
          {{$ctrl.videoLongDesc}}
          Human MCP-2 ProQuantum Immunoassay Kit
          Human MCP-2 ProQuantum Immunoassay Kit
          Human MCP-2 ProQuantum Immunoassay Kit (A355995)
          videoThumbnail
          ►
          Product Specifications
          Analytical sensitivity
          <0.01 pg/mL
          Assay range
          0.0128-5,000 pg/mL
          Sample type/volume
          Plasma
          2-5 µL
          Supernatant
          2-5 µL
          Serum
          2-5 µL
          Hands-on time
          30 min
          Time-to-result
          2 hr
          Homogenous (no wash)
          Yes
          Instrument
          qPCR Instruments
          Product size
          5 x 96 Tests
          Contents
          Assay Dilution Buffer
          Antibody-Conjugate A
          Antibody-Conjugate B
          Antibody-Conjugate Dilution Buffer
          Standard
          Master Mix
          Ligase
          Storage
          -20°C
          Protein name
          CCL8 (MCP2)
          Species (tested)
          Human
          Label or dye
          FAM™
          About This Kit
          The ProQuantum Human MCP-2 Immunoassay Kit is designed to provide quantitative measurements of human MCP-2 protein in small sample volumes. Utilizing proximity ligation assay (PLA) technology, the assay combines the analyte specificity of high-affinity antibody-antigen binding with the signal detection and amplification capabilities of real-time PCR to achieve a simple yet powerful next-generation protein quantitation platform. A user-friendly workflow combined with intuitive Cloud-based software for analytics enables sample-to-answer in just 2 hours.
          • High sensitivity-detect low levels of protein with greater sensitivity than traditional methods
          • Broad dynamic range-≥5 logarithmic units, minimizing sample dilutions to ensure they fall within the range
          • Small sample consumption-use 2-5 µL of sample (compared to 75 µL for triplicate wells with other methods)
          • Fast, easy workflow-2 hours from sample to answer, with no wash steps
          • No proprietary instrument to purchase-runs on any real-time PCR instrument
          ProQuantum immunoassays utilize a matched pair of target-specific antibodies, each conjugated to a DNA oligonucleotide. During antibody-analyte binding, the two DNA oligos are brought into close proximity, which allows for ligation of the two strands and subsequent creation of a template strand for amplification. This platform leverages the sensitivity and large dynamic range of Applied Biosystems TaqMan real-time PCR technology (figure below).
          The assay workflow is fast and easy-2 steps in 2 hours. There are a total of 7 components in each kit (figure below). First, mix the antibody-conjugates, dilute the sample, and create the standard curve in a working plate. Then, using a multi-channel pipette, add the antibody-conjugates and sample (or standard) into the wells of a PCR plate and incubate for 1 hour. Combine the master mix and ligase and add to the wells of the PCR plate, then run the plate on any qPCR instrument. After the run is complete, import the results file into the ProQuantum cloud-based software at https://apps.thermofisher.com/apps/proquantum. Using this software, the data can be analyzed easily to obtain protein concentration values. The software allows you to set up standard curves, design plate layouts, set up customized assay instructions, and obtain robust statistical group-wise comparisons.
          Target information
          This antimicrobial gene is one of several chemokine genes clustered on the q-arm of chromosome 17. Chemokines form a superfamily of secreted proteins involved in immunoregulatory and inflammatory processes. The superfamily is divided into four subfamilies based on the arrangement of N-terminal cysteine residues of the mature peptide. This chemokine is a member of the CC subfamily which is characterized by two adjacent cysteine residues. This cytokine displays chemotactic activity for monocytes, lymphocytes, basophils and eosinophils. By recruiting leukocytes to sites of inflammation this cytokine may contribute to tumor-associated leukocyte infiltration and to the antiviral state against HIV infection.
          Bioinformatics
          Gene aliases : HC14, MCP-2, MCP2, SCYA10, SCYA8
          Gene ID : (Human) 6355
          Gene symbol : CCL8
          Protein Aliases : chemokine (C-C motif) ligand 8, monocyte chemoattractant protein 2, monocyte chemotactic protein 2, small inducible cytokine subfamily A (Cys-Cys), member 8 (monocyte chemotactic protein 2), small-inducible cytokine A8
          UniProt ID (Human) P80075
          Recommended Products
          IL-6 Human Matched Antibody Pair
          Because you are interested in IL-6 Human ELISA Kit.
          SIZE
          10 x 96 tests
          PRICE
          USD 400.00
          Cat # CHC1263
          guarantee_icon
          Performance Guarantee
          If an Invitrogen™ immunoassay doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          Your items have has been added!
          Host server : magellan-search-green-7bf6cff7cf-7xm2d:80/100.66.75.231:80.
          git-commit: cd21911f92a61daaa7859949453b3286702c44a2
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/3.7.0-2026.11.47-1.0