    • See all product categories
  • Applications
    • Bioprocessing
  • Services
    • 360° CDMO and CRO Solutions
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • ELISA Kits ›
          • Caspase 9 ELISA Kits
          Invitrogen
          Human Caspase 9 ELISA Kit
          4 References
          Technical guide & protocol Tech Support
          Technical guide & protocol Tech Support
          Technical guide & protocol Tech Support
          Technical guide & protocol Tech Support
          {{$ctrl.translations['antibody.pdp.skuNotes.additionalInformation']}}:
          {{$ctrl.translations[$ctrl.isSkuNotesCollapsed ? 'antibody.pdp.skuNotes.viewMore' : 'antibody.pdp.skuNotes.viewLess']}}
          Human Caspase 9 ELISA Kit
          Group 53 Created with Sketch.
          Human Caspase 9 ELISA Kit
          Group 53 Created with Sketch.
          FIGURE: 1 / 3
          Human Caspase 9 ELISA Kit
          Representative standard curve for Human Caspase-9 ELISA {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product
          {{$ctrl.videoDesc}}
          {{$ctrl.videoLongDesc}}
          Human Caspase 9 ELISA Kit
          Human Caspase 9 ELISA Kit (BMS2025)
          videoThumbnail
          ►
          Product Specifications
          Analytical sensitivity
          0.4 ng/mL
          Assay range
          1.6-100 ng/mL
          Sample type/volume
          Cell lysate
          50 µL
          Serum
          50 µL
          Supernatant
          50 µL
          Hands-on time
          1 hr 20 min
          Time-to-result
          3 hr 10 min
          Homogenous (no wash)
          No
          Interassay CV
          9%
          Intraassay CV
          6.6%
          Instrument
          Absorbance Microplate Reader
          Product size
          96 Tests
          Contents
          Pre-coated 96 well plate
          Standard
          Sample Diluent
          Assay Buffer concentrate
          Detection Antibody
          anti-rabbit HRP conjugate
          Lysis Buffer
          Wash Buffer
          Chromogen
          Stop Solution
          Adhesive Plate Covers
          Shipping conditions
          Wet ice
          Storage
          2-8°C
          Protein name
          Caspase 9
          Species (tested)
          Human
          Detector antibody conjugate
          HRP
          Label or dye
          HRP
          About This Kit
          The Human Caspase 9 ELISA quantitates Hu Caspase 9 in human serum, cell lysate or cell culture medium. The assay will exclusively recognize both natural and recombinant Hu Caspase 9.
          Principle of the method
          The Human Caspase 9 solid-phase sandwich ELISA (enzyme-linked immunosorbent assay) is designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody has been pre-coated in the wells of the supplied microplate. Samples, standards, or controls are then added into these wells and bind to the immobilized (capture) antibody. The sandwich is formed by the addition of the second (detector) antibody, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen.
          Rigorous validation
          Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity, specificity, precision, and lot-to-lot consistency. See manual for more information on validation.
          Target information
          Caspase 9 (ICE-like apoptotic protease 6, ICE LAP6, apoptotic protease Mch6, apoptotic protease activating factor 3, Apaf3) is a member of the peptidase family C14 that contains a CARD domain. Caspases are cysteine proteases, expressed as inactive precursors, that mediate apoptosis by proteolysis of specific substrates. Caspase 9 is active as a heterotetramer, is present in the cytosol and, upon activation, translocates to the mitochondria. Caspase 9 is involved in the caspase activation cascade responsible for apoptosis execution and cleaves/activates Caspase 3 and Caspase 6. Further, Caspase 9 is involved in the activation cascade of caspases responsible for apoptosis execution. Binding of caspase-9 to Apaf- 1 leads to activation of the protease which then cleaves and activates caspase-3. Caspase 9 promotes DNA damage-induced apoptosis in a ABL1/c-Abl-dependent manner, and proteolytically cleaves poly(ADP- ribose) polymerase (PARP). Caspase 9 is expressed in variety of human tissues. An isoform of rat Caspase-9 has been identified in which the C terminus of full-length Caspase-9 is replaced with an alternative peptide sequence. Mutations in Caspase 9 affect brain development. In mice, insufficient Caspase 9 have resulted in abnormal cerebrum development and neuron development. In humans, dysfunctional Caspase 9 expression vary from tissue to tissue. Low levels of Caspase 9 may play a role in cancer development and neurodegenerative diseases such as Alzheimer's Disease (AD).
          Bioinformatics
          Gene aliases : APAF-3, APAF3, ICE-LAP6, MCH6, PPP1R56
          Gene ID : (Human) 842
          Gene symbol : CASP9
          Protein Aliases : apoptotic protease activating factor 3, apoptotic protease MCH-6, cas9, CASP 9, Caspase9, Caspase-9 precursor, CASPASE-9c, CTD, OTTHUMP00000002324, RNCASP9, RP11-265F14.3, CASP9, caspase 9, apoptosis-related cysteine peptidase, caspase 9b, caspase-9 variant, ICE-like apoptotic protease 6, protein phosphatase 1, regulatory subunit 56, unnamed protein product
          UniProt ID (Human) P55211
          Recommended Products
          IL-6 Human Matched Antibody Pair
          Because you are interested in IL-6 Human ELISA Kit.
          SIZE
          10 x 96 tests
          PRICE
          USD 400.00
          Cat # CHC1263
          guarantee_icon
          Performance Guarantee
          If an Invitrogen™ immunoassay doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          Your items have has been added!
          Host server : magellan-search-green-7bf6cff7cf-d8hkz:80/100.66.77.44:80.
          git-commit: cd21911f92a61daaa7859949453b3286702c44a2
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/3.7.0-2026.11.47-1.0