    • See all product categories
  • Applications
    • Cell Analysis
    • Protein Biology
    • Lab Informatics
    • 360° CDMO and CRO Solutions
  • Help and Support
    • Customer Center
    • Contact Us
    • Certificates of Analysis and Conformance
    • Safety Data Sheets (SDS)
    • Manuals
    • How to Cite Our Products in a Paper
    • Instrument Support
    • Knowledge Base and Product FAQs
            • Services Central
          • ELISA Kits ›
          • TNFR1 ELISA Kits
          Invitrogen
          Human TNFR1 (Soluble-60 kDa) ELISA Kit
          3 References
          View all (5) TNFR1 ELISA Kits
          Technical guide & protocol Tech Support
          Technical guide & protocol Tech Support
          Technical guide & protocol Tech Support
          Technical guide & protocol Tech Support
          {{$ctrl.translations['antibody.pdp.skuNotes.additionalInformation']}}:
          {{$ctrl.translations[$ctrl.isSkuNotesCollapsed ? 'antibody.pdp.skuNotes.viewMore' : 'antibody.pdp.skuNotes.viewLess']}}
          Human TNFR1 (Soluble-60 kDa) ELISA Kit
          Group 53 Created with Sketch.
          Human TNFR1 (Soluble-60 kDa) ELISA Kit
          Group 53 Created with Sketch.
          FIGURE: 1 / 3
          Human TNFR1 (Soluble-60 kDa) ELISA Kit
          Representative standard curve for Human soluble TNF-Receptor I (60kDa) ELISA {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product
          {{$ctrl.videoDesc}}
          {{$ctrl.videoLongDesc}}
          Human TNFR1 (Soluble-60 kDa) ELISA Kit
          Human TNFR1 (Soluble-60 kDa) ELISA Kit (BMS203TEN)
          videoThumbnail
          ►
          Product Specifications
          Analytical sensitivity
          0.05 ng/mL
          Assay range
          0.08-5.0 ng/mL
          Sample type/volume
          Plasma
          10 µL
          Serum
          10 µL
          Supernatant
          10 µL
          Urine
          10 µL
          Hands-on time
          1 hr 20 min
          Time-to-result
          2 hr 10 min
          Homogenous (no wash)
          No
          Interassay CV
          8.6%
          Intraassay CV
          1.9%
          Instrument
          Absorbance Microplate Reader
          Product size
          10 x 96 Tests
          Contents
          Pre-coated 96 well plates
          Standard
          Sample Diluent
          Assay Buffer concentrate
          Detection Antibody Conjugate: anti-rabbit-HRP
          Wash Buffer
          Chromogen
          Stop Solution
          Controls
          Adhesive Plate Covers
          Shipping conditions
          Wet ice
          Storage
          2-8°C
          Protein name
          TNFR1
          Target sub-specificity
          Soluble
          Species (tested)
          Human
          Detector antibody conjugate
          HRP
          Label or dye
          HRP
          About This Kit
          The Human Soluble Tumor Necrosis Factor Receptor 1 (sTNFR1-60 kDa) ELISA quantitates sTNFR1 in human serum, plasma, urine, or cell culture medium. The assay will exclusively recognize both natural and recombinant Hu sTNFR1.
          Principle of the method
          The Human sTNFR1 solid-phase sandwich ELISA (enzyme-linked immunosorbent assay) is designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody has been pre-coated in the wells of the supplied microplate. Samples, standards, or controls are then added into these wells and bind to the immobilized (capture) antibody. The sandwich is formed by the addition of the second (detector) antibody, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen.
          Rigorous validation
          Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity, specificity, precision, and lot-to-lot consistency. See manual for more information on validation.
          Target information
          TNFR1 (Tumor necrosis factor receptor 1) belongs to the tumor necrosis factor superfamily, and is one of the major TNF-alpha receptors. TNFR1 plays an important role in mediating, in cytokine mediated signaling, positive regulation of the NF-Kb pathway, positive regulation of angiogenesis, and negative regulation of gene expression. The extracellular domain of TNFR1 is also released into the circulatory system as soluble TNFR1 (sTNFR1). In humans, the TNFR1 gene is located on chromosome 12. Anti-apoptotic protein BCL2-associated athanogene 4 (BAG4/SODD) and adaptor proteins TRADD and TRAF2 have been shown to interact with TNFR1, and thus play regulatory roles in the signal transduction mediated by the receptor. Germline mutations of the extracellular domains of TNFR1 were found to be associated with the autosomal dominant periodic fever syndrome, and the impaired receptor clearance is thought to be a mechanism of the disease.
          Bioinformatics
          Gene aliases : CD120a, FPF, p55, p55-R, p60, TBP1, TNF-R, TNF-R-I, TNF-R55, TNFAR, TNFR1, TNFR55, TNFR60
          Gene ID : (Human) 7132
          Gene symbol : TNFRSF1A
          Protein Aliases : CD120a antigen, MGC19588, n-smase activation assoc. factor, p62, p63, sCD120a, TNFR, TNFR I, Tumor necrosis factor receptor, soluble CD120a, soluble TNFR1, sTNF RI, sTNFR I, sTNFR1, sTNFRI, TNF alpha gene, TNF-R1, TNF-RI, TNFR-I, TNFR1 beta, tumor necrosis factor binding protein 1, tumor necrosis factor receptor, tumor necrosis factor receptor type 1, tumor necrosis factor-alpha receptor, unnamed protein product
          UniProt ID (Human) P19438
          Recommended Products
          IL-6 Human Matched Antibody Pair
          Because you are interested in IL-6 Human ELISA Kit.
          SIZE
          10 x 96 tests
          PRICE
          USD 400.00
          Cat # CHC1263
          guarantee_icon
          Performance Guarantee
          If an Invitrogen™ immunoassay doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
          • Trademarks
          • Fair Trade Fair Trade
          Korea flag icon
          Korea
          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담
          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat
          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담
          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat
          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담
          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat
          Thermo Fisher Scientific Korea Ltd.
          Representative : Soojin Seok
          Company Registration No. : 117-81-46910
          Thermo Fisher Scientific Solutions LLC
          Representative : Soojin Seok
          Company Registration No. : 114-86-04783
          Location: 12F Suseo Office Building, 281 Gwangpyeong-ro, Gangnam-gu, Seoul, Korea(06349) | Mail-Order Business Registration : 2015-Seoul Gangnam-00898 | Payment : ShinHan Bank 140-004-396660 (Thermo Fisher Scientific Solutions LLC)
          ISMS Logo
          Your items have has been added!
          Host server : magellan-search-blue-7fb9f846c9-dlj7x:80/100.66.76.64:80.
          git-commit: 8207b5afa5c305bd40e0a4f0b9b53fd46f2a8829
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.59.1-2026.09.91-1.0