    • See all product categories
  • Applications
    • Bioprocessing
  • Services
    • 360° CDMO and CRO Solutions
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • ELISA Kits ›
          • ICAM-1 (CD54) ELISA Kits
          The product (cat # {{productObj.productId}}) that you're looking for has been discontinued and replaced with {{productObj.replacementProductId}}. The product (cat # {{productObj.productId}}) that you're looking for has been discontinued. Please contact Technical Support with any questions.
          The product (cat # {{productObj.productId}}) that you're looking for has been discontinued and replaced with {{productObj.replacementProductId}}. The product (cat # {{productObj.productId}}) that you're looking for has been discontinued. Please contact Technical Support with any questions.
          The product (cat # {{productObj.productId}}) that you're looking for will be discontinued once stock is depleted. Please check out our other {{productObj.targetName}} {{productObj.pendingDiscontinuationTargetLabel}} Please check out our other {{productObj.pendingDiscontinuationTargetLabel}} or contact Technical Support with any questions.
          Invitrogen
          Monkey ICAM-1 (Soluble) Matched Antibody Pair
          View all (8) ICAM-1 (CD54) ELISA Kits
          Technical guide & protocol Tech Support
          Technical guide & protocol Tech Support
          Technical guide & protocol Tech Support
          Technical guide & protocol Tech Support
          {{$ctrl.translations['antibody.pdp.skuNotes.additionalInformation']}}:
          {{$ctrl.translations[$ctrl.isSkuNotesCollapsed ? 'antibody.pdp.skuNotes.viewMore' : 'antibody.pdp.skuNotes.viewLess']}}
          Monkey ICAM-1 (Soluble) Matched Antibody Pair (BMS648MST)
          Group 53 Created with Sketch.
          Monkey ICAM-1 (Soluble) Matched Antibody Pair (BMS648MST)
          Group 53 Created with Sketch.
          FIGURE: 1 / 2
          Monkey ICAM-1 (Soluble) Matched Antibody Pair
          Matched Antibody Pair {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product
          {{$ctrl.videoDesc}}
          {{$ctrl.videoLongDesc}}
          Monkey ICAM-1 (Soluble) Matched Antibody Pair (BMS648MST)
          videoThumbnail
          ►
          Product Specifications
          Analytical sensitivity
          6.3 U/mL
          Assay range
          6.3-100 U/mL
          Sample type/volume
          Plasma
          10 µL
          Serum
          10 µL
          Supernatant
          10 µL
          Hands-on time
          24 hr
          Time-to-result
          1 hr 10 min
          Homogenous (no wash)
          No
          Instrument
          Absorbance Microplate Reader
          Product size
          10 x 96 Tests
          Contents
          Capture (coating) Antibody
          Detection Antibody
          Standard
          Sample Diluent
          Shipping conditions
          Wet ice
          Storage
          -20°C
          Protein name
          ICAM-1 (CD54)
          Target site-specificity
          Target sub-specificity
          Soluble
          Detector antibody conjugate
          HRP
          Label or dye
          HRP
          About This Kit
          The Monkey ICAM-1 Matched Antibody Pair Kit comes with pre-matched antibody pairs, standards, and sample diluent to develop your own enzyme linked immunosorbent assays (ELISA) to detect and quantify protein levels of moneky ICAM-1. Buffer reagents and microplates needed to complete the ELISA reaction are sold separately.
          Principle of the method
          ELISAs are designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody is coated to the bottom of the wells of a microplate, which is an overnight process. Samples, standards, or controls are then added into these wells and bind to the immobilized (capture) antibody. A sandwich is formed by the addition of the second (detector) antibody, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen.
          Target information
          ICAM-1 (CD54) is an 85-110 kDa single-chain type 1 integral membrane glycoprotein with an extracellular domain of five immunoglobulin superfamily repeats, a transmembrane region and a cytoplasmic domain. ICAM-1 has 7 potential N-linked glycosylation sites and shares considerable amino acid sequence homology with ICAM-3 (CD50) and ICAM-2 (CD102). ICAM-1 binds to integrins of type CD11a/CD18 (leukocyte adhesion molecule, LFA-1), or CD11b/CD18 (Mac-1) and is exploited by Rhinovirus as a receptor. ICAM-1 is expressed by activated endothelial cells and detected on epithelial cells, fibroblasts, chondrocytes, B lymphocytes, T lymphocytes (low), monocytes, macrophages, dendritic cells and neutrophils, with lower levels that increase upon inflammation. ICAM-1 is also detected in some carcinoma and melanoma cells. Soluble ICAM-1 is detectable in the plasma and is elevated in patients with various inflammatory syndromes.
          Bioinformatics
          Protein Aliases : ICAM-1
          Recommended Products
          IL-6 Human Matched Antibody Pair
          Because you are interested in IL-6 Human ELISA Kit.
          SIZE
          10 x 96 tests
          PRICE
          USD 400.00
          Cat # CHC1263
          guarantee_icon
          Performance Guarantee
          If an Invitrogen™ immunoassay doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          Your items have has been added!
          Host server : magellan-search-green-7bf6cff7cf-xbl4w:80/100.66.75.228:80.
          git-commit: cd21911f92a61daaa7859949453b3286702c44a2
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/3.7.0-2026.11.47-1.0