Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • ELISA Kits ›
          • ERK1/2 ELISA Kits

          Invitrogen

          Multispecies ERK1/ERK2 (Total/Phospho) InstantOne™ ELISA Kit

          4 References
          View all (4) ERK1/2 ELISA Kits

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Technical guide & protocol Tech Support
          Technical guide & protocol Tech Support
          Technical guide & protocol Tech Support
          Technical guide & protocol Tech Support
          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          Multispecies ERK1/ERK2 (Total/Phospho) InstantOne™ ELISA Kit
          Group 53 Created with Sketch.
          Multispecies ERK1/ERK2 (Total/Phospho) InstantOne™ ELISA Kit
          Group 53 Created with Sketch.

          FIGURE: 1 / 3

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          Multispecies ERK1/ERK2 (Total/Phospho) InstantOne™ ELISA Kit

          Representative data for ERK1/ERK2 (Total/Phospho) Multispecies InstantOne™ ELISA. A431 cells were pre-treated with 10 µM AG1478 (an EGFR kinase inhibitor) for 3 hours (-) or treated with 10 ng/mL EGF for 10 min (+). Cells were analyzed for total (left) or phosphorylated (right) ERK 1/2 by InstantOne™ ELISA. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          Multispecies ERK1/ERK2 (Total/Phospho) InstantOne™ ELISA Kit
          Multispecies ERK1/ERK2 (Total/Phospho) InstantOne™ ELISA Kit (85-86013-11)
          videoThumbnail
          ►

          Product Specifications

          Sample type/volume

          Cell Lysate
          20 µL

          Hands-on time

          1 hr 20 min

          Time-to-result

          1 hr 10 min

          Homogenous (no wash)

          No

          Instrument

          Absorbance Microplate Reader

          Product size

          96 Tests

          Contents

          Capture Antibody
          Detection Antibody Reagent
          InstantOne™ Assay Plate
          Cell Lysis Buffer
          Enhancer Solution
          Wash Buffer (10X)
          Substrate Reagent (TMB)
          Stop Solution
          Positive Control Cell Lysate (lyophilized)
          Plate seal

          Shipping conditions

          Wet ice

          Storage

          2-8°C

          Protein name

          ERK1/2

          Target sub-specificity

          Total

          Species (tested)

          Human

          Assay kit format

          InstantOne ELISA

          Detector antibody conjugate

          Biotin

          Label or dye

          HRP

          About This Kit

          The InstantOne™ ELISA is specifically engineered for accurate measurement of total and phosphorylated human, mouse, and hamster ERK 1/2 in cell lysates. Detection of rat ERK 1/2 is expected. The InstantOne™ ELISA kit allows for fast analysis of samples in approximately one hour. All reagents used in a traditional sandwich ELISA are added in solution to a plate followed by a wash step and detection with the TMB colorimetric substrate.

          Target information

          ERK1 and ERK2 are widely expressed and are involved in the regulation of meiosis, mitosis, and postmitotic functions in differentiated cells. Many different stimuli, including growth factors, cytokines, virus infection, ligands for heterotrimeric guanine nucleotide-binding protein (G protein)-coupled receptors and transforming agents, activate the ERK1 and ERK2 pathways. When growth factors bind to the receptor tyrosine kinase, Ras interacts with Raf, the serine/threonine protein kinase and activates it as well. Once actived, Raf phosphorylates serine residue in 2 further kinases, MEK1/2, which in turn phosphorylates tyrosine/threonine in extracellular-signal regulated kinase (ERK) 1/2. Upon activation, the ERKs either phosphorylate a number of cytoplasmic targets or migrate to the nucleus, where they phosphorylate and activate a number of transcription factors such as c-Fos and Elk-1.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          Bioinformatics

          Gene aliases : ERK, ERK-1, ERK-2, ERK1, ERK2, ERT1, ERT2, HS44KDAP, HUMKER1A, MAPK1, MAPK2, MAPK3, NS13, p38, p40, p41, p41mapk, p42-MAPK, P42MAPK, p44-ERK1, p44-MAPK, P44ERK1, P44MAPK, PRKM1, PRKM2, PRKM3

          Gene ID : (Human) 5594, (Human) 5595

          Gene symbol : MAPK1, MAPK3

          Protein Aliases : ERK-1, ERK-2, erk1 erk2, ERK1b, p42, p42 MAP Kinase, Erk1, cancer susceptibility protein, ERK1c, Erk2, ERT1, ERT2, Extracellular signal-regulated kinase 1, Extracellular signal-regulated kinase 2, extracellular signal-related kinase 1, Insulin-stimulated MAP2 kinase, MAP kinase 1, MAP kinase 2, MAP kinase 3, MAP kinase isoform p42, MAP kinase isoform p44, MAPK 1, MAPK 2, Microtubule-associated protein 2 kinase, Mitogen-activated protein kinase 1, Mitogen-activated protein kinase 2, Mitogen-activated protein kinase 3, p42-MAPK, p44-ERK1, p44-MAPK, protein tyrosine kinase ERK2, unnamed protein product

          UniProt ID (Human) P28482, (Human) P27361

          Recommended Products

          IL-6 Human Matched Antibody Pair

          Because you are interested in IL-6 Human ELISA Kit.

          SIZE

          10 x 96 tests

          PRICE

          USD 400.00

          Cat # CHC1263

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ immunoassay doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-744bc48644-fvx7n:80/100.66.78.247:80.
          git-commit: 747bde55a712f6f97bf7760408d445eefba4e16f
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.48.0-Offline