Vector NTI Software
Curate. Discover. Design. Confirm.
Invitrogen Vector NTI software is a completely integrated suite of sequence analysis and design tools that help you manage, view, analyze, transform, share, and publish diverse types of molecular biology data, all within one graphically rich analysis environment.
- Curate—store and manage collections, visualize maps, and search sequences
- Discover—analyze, compare, and contrast sequences
- Design—cloning strategy, primers for PCR, cloning, and resequencing, and gel simulations of sequences
- Confirm—contig assembly, sequence validation, and literature validation
Vector NTI Express Designer Software
![]() |
Vector NTI Express Designer Software leads the way in engineering the next generation of in silico cloning and assembly. The newly developed interface is a virtual project canvas for both simplified drag-and-drop vector construction and simplified sequence alignment between projects. Vector NTI Express Designer Software also provides access to Invitrogen GeneArt web tools to obtain optimized protein expression levels. Learn more or download a trial. |
Explore our latest platform, Vector NTI Express Designer Software—now available for Mac or PC operating systems.
Vector NTI® Express Software
![]() |
Vector NTI® Express Software retains the trusted tools of Vector NTI® Advance Software with a brand-new, faster interface. Since your Vector NTI® Express license works with Vector NTI® Advance Software as well, think of a Vector NTI® Express Software purchase as an investment—you can stay with Vector NTI® Advance Software as long as you’d like, and Vector NTI® Express Software will be available to you whenever you’re ready. In addition to boasting a new interface, Vector NTI® Express Software also has dedicated Mac® OS X® support, and new bioinformatics tools from Life Technologies will be delivered seamlessly through automatic updates to the existing software. Learn more or download a trial. |
Explore Vector NTI® Express Software — now available for Mac® or PC operating systems.
Interested in Vector NTI Advance Software?
|
Vector NTI Advance software is a multifunctional desktop sequence analysis application suite that provides integration, data management, and scalability. The software is cited in more than 20,000 publications and is compatible with CE sequencing. Version 11.5, launched in 2010, is compatible with Windows 7 and Mac OS X running Windows under Parallels Desktop 5 or Boot Camp. New features in Version 11.5 include:
|
Which Vector NTI Software is right for you?
We understand that your lab has specific needs. We are happy to provide a custom quote for Vector NTI Express or Vector NTI Advance Software.
NEW Invitrogen Anza Restriction Enzymes for Uploading to Vector NTI
| Instructions for Importing Anza Restriction Enzymes Vector NTI Express or Vector NTI Express Designer Vector NTI Advance |
Data File for Importing Anza Restriction Enzyme Data File |
Contact us
Email our software specialists at bioinfosales@lifetech.com for more information, or if you’d like assistance in determining the best software and license agreement to meet your needs.