Search
We are dedicated to your success. Get back on track. View our expert recommendations for commonly encountered problem scenarios.
View the relevant questions below:
Beginning your experiment? Visit our
Our biochemistry has been tailored to minimize the amplification of rRNA. We cannot disclose the specific mechanisms that are used. Additionally, cross-hybridization signal is reduced through our development of assay conditions that take advantage of the thermodynamic differences between labeled RNA targets and labeled cDNA targets that are hybridized to microarrays with attached DNA probes.
The HT HWS reagents have been rigorously tested and validated to perform to our system specifications. Data from GeneChip™ HT Array Plates processed with HT HWS Kit reagents showed similar data and results to HT Array Plates processed with manually prepared reagents.
For Research Use Only. Not for use in diagnostic procedures.