Search
Whether you are interested in detecting polymicrobial infections, slow-growing anaerobic species, or antibiotic resistance (ABR) genes, we have an extensive list available for your research needs.
Bacterial species |
|
|---|---|
Acinetobacter baumannii |
Peptoniphilus asaccharolyticus |
Bacteroides fragilis |
Peptoniphilus harei |
Citrobacter koseri |
Peptoniphilus ivorii |
Citrobacter freundii |
Peptostreptococcus anaerobius |
Citrobacter braakii |
Proteus mirabilis |
Clostridium perfringens |
Proteus vulgaris |
Clostridium septicum |
Providencia stuartii |
Clostridium sordellii |
Pseudomonas aeruginosa |
Clostridium novyi A,B |
Serratia marcescens |
Clostridium histolyticum |
Staphylococcus aureus |
Corynebacterium striatum |
Staphylococcus epidermidis |
Enterobacter cloacae |
Staphylococcus haemolyticus |
Enterococcus faecalis |
Staphylococcus lugdunensis |
Enterococus faecium |
Stenotrophomonas maltophilia |
Escherichia coli |
Streptococcus agalactiae |
Finegoldia magna |
Streptococcus dysgalactiae |
Klebsiella pneumoniae |
Streptococcus anginosus |
Klebsiella oxytoca |
Streptococcus pyogenes |
Streptococcus pyogenes |
|
Morganella morganii |
|
Fungal species |
|
|---|---|
Candida auris |
Candida albicans |
Candida glabrata |
Candida parapsilosis |
Candida krusei |
Candida tropicalis |
Antibiotic class |
Gene family |
|---|---|
Extended-spectrum beta-lactamases |
blaGES |
blaVEB |
|
blaPER |
|
Extended-spectrum beta-lactamases |
blaSHV |
Extended-spectrum beta-lactamases |
blaCTX-M |
Carbapenemase |
blaOXA-51 |
blaOXA-23 |
|
blaOXA-2 |
|
blaOXA-1 |
|
Carbapenemase
|
blaOXA-48 |
blaKPC |
|
blaVIM |
|
blaNDM |
|
Carbapenemase |
blaIMP |
AmpC beta-lactamase |
blaACC / blaFOX |
AmpC beta-lactamase |
blaACT / blaMIR |
AmpC beta-lactamase |
blaDHA |
blaCMY/blaLAT |
|
blaMOX/blaCMY |
|
Beta-Lactamase |
blaTEM |
Lincosamide, Macrolide, Streptogramin |
Cfr |
Trimethoprim |
dfrA1 / A5 / A12 / A17 |
Trimethoprim |
dfrA14 |
dfrB1 / B5 |
|
dfrG |
|
Macrolides |
mef(A) |
ere(B) |
|
mph (A) |
|
Macrolides |
msr (A) |
Macrolides |
erm(A) / (B) / (C) |
Methicillin |
mecA / mecC |
Sulfonamide |
sul1 / sul2 |
Colistin |
mcr-1 / 2 / 3 |
Tetracycline |
tet(M) |
Tetracycline |
tet(A) / (B) / (S) |
Tetracycline |
tet(M) / (A) / (B) / (S) |
Vancomycin |
vanA / vanB |
Nitromidazole |
nimB / D / J / E |
Quinolone |
qnrB |
Quinolone |
qnrA / qnrS |
Aminoglycoside |
aac(3)-Ia |
aac(3)-Ib/aac(6')-Ib'' |
|
aac(6')-30/aac(6')-Ib |
|
aac(6')-Ib |
|
aac(6')-Ib' |
|
aac(6')-Ib11 |
|
ant(3'')-Ih/aac(6')-IId |
|
aac(6')-Ib-cr |
|
aadA1 / A12 / A15 / A22 |
The species listed below are known to contain one or more of the previously listed ABR genes.
|
Urinary research |
|---|
Pseudomonas aeruginosa |
Acinetobacter baumannii |
Klebsiella sp. |
Staphylococcus aureus |
Streptococcus sp. |
Serratia marcescens |
Citrobacter freundii |
Proteus sp. |
Providencia stuartii |
Enterococcus sp. |
Escherichia coli |
Ureaplasma urealyticum |
|
Women’s health / STI research |
|---|
Escherichia coli |
Chlamydia trachomatis |
Neisseria gonorrhoeae |
Staphylococcus aureus |
Atopobium vaginae |
Bacterial Vaginosis-Associated Bacterium 2 |
Enterococcus faecalis |
Mobiluncus curtisii |
Mobiluncus mulieris |
|
Respiratory research |
Klebsiella pneumoniae |
Staphylococcus aureus |
Streptococcus pneumoniae |
|
Gastrointestinal research |
|---|
Escherichia coli |
Salmonella |
Shigella |
Listeria monocytogenes |
Vibrio sp. |
For Research Use Only. Not for use in diagnostic procedures.