Search
One Lambda MagSort addresses emerging interests and challenges in adsorption and elution techniques for HLA antibody testing. Targeting individual HLA antigens enables a more focused dataset, to explore and define complex HLA antibody profiles at the antigen-specific level. This novel technology streamlines the process, reducing hands-on time for the laboratory staff, and standardizes a unique isolation approach compared to more cumbersome cell-based approaches.
| MagSort Class II HLA-DRB1 | MagSort Class II HLA-DQ | MagSort Class II HLA-DP | |||
A*01:01 |
B*07:02 |
C*01:02 |
DRB1*01:01 | DQB1*02:01 DQA1*02:01 |
DPB1*01:01 DPA1*01:03 |
A*02:01 |
B*08:01 |
C*02:02 |
DRB1*03:01 |
DQB1*02:02 DQA1*02:01 |
DPB1*02:01 DPA1*01:03 |
A*03:01 |
B*13:01 |
C*03:03 |
DRB1*04:01 |
DQB1*03:01 DQA1*02:01 |
DPB1*03:01 DPA1*01:03 |
A*11:01 |
B*15:01 |
C*03:04 |
DRB1*04:04 |
DQB1*03:02 DQA1*02:01 |
DPB1*04:01 DPA1*01:03 |
A*24:02 |
B*44:02 |
C*04:01 |
DRB1*07:01 |
DQB1*03:03 DQA1*02:01 |
DPB1*04;02 DPA1*01:03 |
A*25:01 |
B*46:01 |
C*05:01 |
DRB1*08:01 |
DQB1*04:01 DQA1*02:01 |
DPB1*11:01 DPA1*01:03 |
| A*29:01 | B*53:01 | C*07:02 | DRB1*11:01 | DQB1*05:01 DQA1*01:01 |
DPB1*17:01 DPA1*02:01 |
| A*30:01 | B*57:01 | C*12:03 | DRB1*12:01 | DQB1*06:02 DQA1*01:01 |
DPB1*19:01 DPA1*01:03 |
| A*33:03 | B*67:01 | C*15:02 | DRB1*13:01 | DQB1*06:03 DQA1*01:03 |
|
| A*68:01 | B*73:01 | C*16:01 | DRB1*15:01 | ||
| C*17:01 | DRB1*16:01 |
For research use or further manufacturing. Not for diagnostic use or direct administration into humans or animals.