On this page:
Take control of your results with versatility and actionable insights
AccuSEQ real-time PCR software is a versatile tool designed to enhance your analytical capabilities across a range of applications. Compatible with MycoSEQ, ResDNASEQ, SteriSEQ, and ViralSEQ kits as well as custom assays, AccuSEQ software offers a unified platform for detecting impurities, identifying contamination, and quantifying titers. Its robust data analysis and reporting features help ensure precise and actionable insights, making it a critical component of your testing workflow. Whether you're working with standard assays or developing custom solutions, AccuSEQ software offers the reliability and flexibility needed to meet the highest standards of quality and compliance.
Mycoplasma detection
For rapid and accurate mycoplasma detection, AccuSEQ software offers a plug-in analysis module for automated presence/absence calls when using the MycoSEQ mycoplasma detection system.
Key features and benefits: 
  • Optimized and validated for the MycoSEQ Mycoplasma Detection assay
  • Automated detection of PCR inhibition in sample wells
  • Automated detection of discriminatory positive control (DPC)
  • Robust data analysis parameters for the MycoSEQ Mycoplasma Detection assay
  • Flexibility for experiments—call settings may also be modified to meet the specifications of individual laboratories
AccuSEQ software v4.0 shows the automated presence/absence calls of mycoplasma contamination.
AccuSEQ software v4.0 shows the MycoSEQ results analysis table where calls are made based on the three acceptance criteria–C, T, and derivative value of test samples and inhibition controls.
Residual host cell DNA analysis
AccuSEQ software offers factory default templates to facilitate experiment set up for the accurate and sensitive quantitation of host cell DNA impurities in both the purification process and in drug substance samples. 
Key features and benefits: 
  • Streamlined resDNASEQ experiment setup (using factory default templates) and data reporting 
  • Automatic calculations of the resDNASEQ standard curve with curve fitting parameters 
  • Accurate quantitation of samples with % CV 
  • Computation of key metrics including % recovery, total DNA in pg/mL, and pg DNA/mg protein 
  • Flexibility for custom experiments to meet the specifications of individual laboratories 
  • Customizable output layout that can be exported to a variety of formats 
  • Comprehensive reports which help eliminate the need for Excel sheets for calculations and preserve the audit trail 
For plate setup using AccuSEQ software, simply enter sample names, serial dilution factor, and spike-in information.
After the experiment run, AccuSEQ software v4.0 automatically sets up the standard curve calculations and creates a visual display of the results with relevant curve fitting parameters, such as slope, intercept and efficiency, and R2.
AccuSEQ software v4.0 shows the standard curve plot with samples overlayed and the results analysis tab on the right.
The resDNASEQ results table in AccuSEQ software v4.0 shows key metrics including Quantity Mean, Quantity % CV, % Recovery, Total DNA per mL, and Total DNA per Protein Concentration.
AccuSEQ software v4.0 shows the Print report section where users can customize report contents.
Snapshot of the resDNASEQ Result Summary from the PDF report generated by AccuSEQ software v4.0—the comprehensive summary includes all important metrics required in the resDNASEQ experiment, such as % CV, % Recovery, and Total DNA per mL (dilution adjusted).
Rapid sterility testing
For rapid and accurate sterility testing, the AccuSEQ Software offers presence or absence calls when using the SteriSEQ Rapid Sterility Testing Kit, allowing for easy analysis.
Key features and benefits: 
  • Automated analysis with presence/absence calls 
  • Bulk edit feature to allow for custom rule edits 
  • Discriminatory positive control to help eliminate false positives 
  • Predefined templates to allow for streamlined experiment setup 
  • Comprehensive reports which help eliminate the need for Excel sheets for calculations and preserve the audit trail 
AccuSEQ software v4.0 shows the automated presence/absence calls of sterility testing.
ViralSEQ information
Detection of mouse minute virus (MMV) and vesivirus
For rapid and accurate MMV and vesivirus detection, the AccuSEQ Software offers: 
  • A robust, optimized, and validated system offering data analysis designed for use with ViralSEQ detection systems
  • Automated detection for rapid assessment of PCR inhibition in sample wells
  • Analysis options to identify true positives from false positives with discriminatory positive control
  • Flexible call settings to help you meet specifications for unique lab requirements
Quantitation of Sf-rhabdovirus with AccuSEQ Software 
The AccuSEQ Software offers factory streamlined experimental set up for the accurate and sensitive quantitation of Sf-rhabodvirus contamination in both the purification process and in drug substance samples. 
Key features and benefits: 
  • Streamlined experiment setup with factory default templates for easy data reporting
  • Automated calculations to help eliminate the need for spreadsheets and preserve the audit trail
  • Comprehensive reports that present key metrics, including % Recovery, Total RNA in pg/mL, and pg RNA/mg Protein
  • Flexibility to easily adjust for custom experiments and meet specialized specifications and reporting needs
Custom experiments
The custom experiment mode—“Create New”—of the AccuSEQ software enables you to create custom experiment setups. It guides you through the experiment properties, qPCR run method details, and plate setup where there are easy-to-use tools for defining and setting up standards, controls, and unknowns. This custom workflow also supports ViralSEQ MMV, vesivirus, and resDNASEQ Sf9-Baculovirus duplex assays.
AccuSEQ software v4.0 shows the custom—“Create New”—selection where users have the ability to create an experiment with their own specific and unique parameters.
AccuSEQ software v4.0 shows the custom—“Create New”—selection where users have the ability to create an experiment differing from the default experiment templates. You can choose the experiment type, reagent type, and ramp speed.
Example of AccuSEQ software v4.0 module for ViralSEQ MMV assay demonstrating the amplification readout.
Two views of customizable export sample setup and results with capability to group by well or sample in AccuSEQ software v4.0.
Customized output for SEQ Analytical Assays
AccuSEQ software is designed to create customized output for SEQ assays, helping you export complete, traceable and simple outputs, helping to streamline the analysis process. Rather than outputting Ct and requiring further downstream calculations, the customized output feature allows you to have a specialized output to match your needs. For example, the LentiQuant physical titer outputs the viral particles/mL that help determine the titer of lentivirus in viral supernatants and purified viruses.
Assay
Specialized output
ViralSEQ MMV detection kit
  • Call
  • Call assessment 
ViralSEQ Vesivirus detection kit 
  • Call
  • Call assessment 
MycoSEQ Plus mycoplasma detection kit 
  • Call
  • Call assessment 
SteriSEQ detection kit 
  • Call
  • Call assessment
All resDNASEQ Quantitative Kits
  • All resDNASEQ Quantitative Kits Ct, Ct mean
  • Qty, Qty mean, Qty % CV
  • Dilution adjusted
  • % Recovery
resDNASEQ Quantitative E1A DNA Fragment Length Kits
  • All resDNASEQ Quantitative Kits Ct, Ct mean
  • Qty, Qty mean, Qty % CV
  • Dilution adjusted
  • % Recovery
resDNASEQ Quantitative Plasmid DNA - Kanamycin Resistance Gene Kits and ViralSEQ Quantitative Sf-Rhabdovirus Kit 
  • % Recovery 
  • Mass (g) 
ViralSEQ Lentivirus Physical Titer Kit 
  • Viral particles per mL (VP/mL) 
ViralSEQ Lentivirus Proviral DNA Titer Kit 
  • Provirus copies per cell 
Customized output for quantitative Sf9 and Baculovirus DNA kit using the AccuSEQ software v4.0
Customized output for Lentivirus physical titer kit using AccuSEQ software v4.0
Analytics Knowledge Hub
Explore the Analytics Knowledge Hub and discover a robust collection of webinars, articles, eBooks, infographics, and other digital resources to help you optimize analytical testing in your bioprocess workflow.
Request a technical consultation
No matter your questions, we are here to help. Our experienced team of scientists and technical specialists is ready to assist you in choosing the right solution and products. Complete this form to start the conversation.
Additional bioprocessing resources
Discover areas where you can cut costs and gain efficiency using our integrated calculator.
Explore our collection of information and insights to support your drug development journey.
Learn, evaluate, connect.
Connect with a Bioprocessing representative or request more info.
For research use or further manufacturing. Not for diagnostic use or direct administration into humans or animals.