Search
MagicMedia™ E. coli Expression Medium is an auto induction growth media used with T7-regulated E. coli strains that facilitates protein expression without the addition of IPTG. Life Technologies offers MagicMedia™ E. coli Expression Medium available in both SoluPouch™ powder and ready-to-use liquid formats for convenience and ease of use.