Shaping the future of health together
The Applied Biosystems GeneTitan MC Fast Scan Instrument provides the highest throughput and laboratory productivity per technician by minimizing user intervention and allowing unattended, overnight processing of large numbers of samples in parallel. This unique degree of hands-off automation means your data will be consistently reproducible while maximizing productivity.
Product overview
GeneTitan MC Fast Scan Instrument
GeneTitan image
The GeneTitan MC Fast Scan Instrument and proven Axiom array plates in 96- and 384-array layouts are the first to provide a hands-free, automated solution for genome-wide SNP genotyping.
The instrument automates array processing from target hybridization to data generation by combining a hybridization oven, fluidics processing, and state-of-the art imaging device into a single bench-top instrument.
The imaging device in the GeneTitan MC Fast Scan Instrument uses an external, high-intensity xenon lamp and dual excitation and emission filters to capture images from array plates for use in genotyping and microbiome studies.
Key benefits include:
  • Fast—condenses hands-on processing time to as little as 30 minutes, images an array in less than five minutes, and operates unattended overnight
  • Scalable—meets both medium- and high-throughput needs, enables fast time to data, and requires minimal manual intervention
  • Flexible—supports genotyping studies on multiple array plate formats
  • Accurate—enables high-quality, consistent data by processing multiple samples under identical conditions
  • Adaptable—creates flexible workflows and sample registration via GeneChip Command Console Software
GeneChip Command Console Software
GeneChip Command Console Software provides customization, automation, and integration flexibility along with full-range support:
  • Provides summary of probe cell intensity data (CEL file generation); enables sample and array registration, data management, instrument control, and automatic and manual image gridding
  • Enables flexiblility to customize, automate, and integrate workflows with a variety of laboratory information management systems
  • Supports the full range of assays by enabling seamless integration with downstream primary analysis applications such as Axiom Analysis Suite and Transcriptome Analysis Console (TAC) Software
GeneChip Command Console Software logo
Product specifications
GeneTital MC Fast Scan Instrument
Dimensions
55” W x 33” D x 26” H (139.7 cm W x 83.82 cm D x 66 cm H)
Applications
Microarray processing
Weight
147 kg