Search
Microarrays have revolutionized genomic research by providing a high-throughput, scalable platform for analyzing gene expression and genetic variation across thousands of genes simultaneously. With their versatility, efficiency, and reliability, microarrays remain a cornerstone of modern genomics—driving innovation in healthcare, agriculture, and basic research.
To further advance innovation and better serve our customers, we are proud to launch the new Applied Biosystems™ SwiftArrayStudio™ Microarray Analyzer. This next-generation instrument helps meet the growing demands for speed in high-throughput microarray analyses with a streamlined two-day workflow and reduced hands-on time, making it easier and faster to achieve results. The instrument is designed to deliver outstanding performance, flexibility, and impact across a broad range of real-world applications.
| Two-day (30-hour) turnaround time | Minimal touchpoints—load arrays and reagents and walk away |
| High throughput—process up to 16 plates (6,144 samples) in a 5-day work week | Integrated user interface with command console—no dock station required to perform instrument operations |
| Fast—scan each sample in as little as 15 seconds to get your genotype results the next day | Scalable—run one sample plate at a time or scale up to 4X sample plates (*various workflow options are available to increase throughput in addition to batching) |
| One instrument, automated array processing— hybridize, stain, wash, and scan | Robust—durable interior mechanics for increased uptime |
The SwiftArrayStudio Microarray Analyzer has incorporated internal features that improve the speed at which array plates are processed. These changes increase the speed at which data can be collected from samples.
Thermo Fisher Scientific offers a growing number of microarrays in Applied Biosystems Axiom array plate format. The SwiftArrayStudio Microarray Analyzer has been designed to be able to run all current and future array plates, regardless of content, on the array. These include:
Many of the key features of the new system advance usability and efficiency. These features make setting up, using, and maintaining the system easy.
The SwiftArrayStudio Microarray Analyzer has been designed to provide a robust and reliable platform that supports laboratory uptime. In addition to helping keep the data flowing, this helps minimize service visits needed.
The SwiftArrayStudio Microarray Analyzer offers a robust and efficient platform for a variety of genotyping applications.
The SwiftArrayStudio Microarray Analyzer was designed to be easy to use and easy to service when needed. These designs take advantage of the Thermo Fisher Scientific global service and support teams so you can quickly get back to your research.
Attribute |
Specification |
| Dimensions (W x H x D) | 54.52 x 25.03 x 26.92 in. (138.48 x 68.58 x 68.39 cm) External waste bottle: 6 x 10 x 8 in. (15.24 x 25.4 x 20.32 cm) |
| Weight | 375 lb (170.1 kg) |
| Temperature range | 15°C–30°C (59°F–85°F) |
| Humidity range | 20%–80% relative humidity, Noncondensing |
| Pollution degree | Pollution degree 2: Install the instrument in an environment that normally only has nonconductive pollutants, such as dust particles or wood chips. Typical environments are laboratories and sales and commercial areas. |
| Operating altitude | Located between sea level and 6,500 ft. (2,000 m) above sea level |
| Electrical supply | 100–240 VAC |
| Current | 6.2–2.6 A |
| Main supply voltage fluctuations | ±10% VAC |
| Illumination | LED light source |
| Pixelation | 4.1 megapixels |
| Scan time, 96-array plate | 105 minutes |
| Scan time, 384-array plate | 95 minutes |
| Configuration | Scan time entire plate | Scan time per sample | Max samples per week* |
| 96-array plate | 105 minutes | 65.6 seconds | 1,536 |
| 384-array plate | 95 minutes | 15 seconds | 6,144 |
*Assumes a five-day work week.
For Research Use Only. Not for use in diagnostic procedures.