A complete, integrated research solution for next-day results
The Applied Biosystems SwiftArrayStudio solution integrates the SwiftArrayStudio Microarray Analyzer, Axiom SwiftArray Assay, and high-performance Axiom arrays to deliver fast, reliable, and scalable genotyping. The fully integrated analyzer automates array processing, reducing hands-on time, while the simplified Axiom SwiftArray Assay enables sample prep to hybridization readiness in one day. With flexible catalog and custom Axiom array designs, the solution supports human and agrigenomic research and enables results in just two days—accelerating the path from DNA to actionable insights.
Highlights
  • Next-day genotyping results—Sample-to-report turnaround in as little as 30 hours to support faster research timelines.
  • High-throughput microarray processing—Process up to 16 plates (1,536 samples) in a standard 5-day work week using a 96-well format.
  • Automated array processing in a single instrument—Integrated hybridization, washing, staining, and scanning help reduce manual steps after target preparation.
  • Designed to minimize hands-on time—Load arrays and reagents, then walk away — supporting consistent processing and improved reproducibility.
  • Fast, high-resolution scanning—Scan a single sample in as little as 15 seconds.
  • Intuitive, integrated user interface—Built-in touchscreen, streamlining instrument operation without the need for a separate dock station.
  • Flexible and scalable workflows—Run one plate at a time or scale up to multiple plates with workflow options that support batching and throughput needs.
  • Durable design for reliable operation—Robust internal mechanics are engineered to support uptime in high-throughput research environments.
Speed
Faster array processing and data acquisition
The SwiftArrayStudio Microarray Analyzer is designed to accelerate array processing and data collection while supporting consistent, high-quality results.
Walk-away array processing
A single plate can be processed without manual intervention in approximately 21 hours after target preparation.
Optimized scanning performance
Advanced autofocus algorithms during scanning support efficient signal acquisition and optimal image quality.
High-throughput scan times
Average scan time for a full 96-well plate is ~105 minutes (~65 seconds per sample) and scan time for a full 384-well plate is ~95 minutes (~15 seconds per sample).
Flexibility
One platform. Multiple array applications.
SwiftArrayStudio is designed to support a range of Applied Biosystems Axiom array formats, enabling laboratories to run diverse genotyping applications on a single microarray analyzer.
PangenomePro arrays
Designed for population genomics and large-scale research, enabling comprehensive genome-wide variant analysis across diverse populations. Utilize PangenomePro Plus to unlock even more pharmacogenomics genes.
PharmacoPro arrays
Optimized for pharmacogenomics research, supporting studies of genetic variation relevant to drug response, metabolism, and safety.
Custom Axiom arrays
Available in 24-, 96-, and 384-array formats, allowing researchers to tailor content for specific research questions and study designs.
(Array content customization supported via MRSL myDesign.)
Future agrigenomics support
SwiftArrayStudio is architected to support additional research applications over time, including agrigenomics arrays such as planned bovine-focused content, with more applications expected in the future.
Note: Agrigenomics offerings are under development; availability will vary by assay.
Efficiency
Designed to maximize throughput and usability
Key system features help laboratories run efficiently while minimizing manual effort and operational complexity.
High-throughput capacity
Process up to 16 plates (1,536 samples) in a standard 5-day work week using a 96-well format.
Automated array processing
Integrated hybridization, washing, staining, and scanning streamline array workflows after target preparation.
Minimal hands-on interaction
Load arrays and reagents, then walk away—helping reduce operator time and variability.
Scalable workflows
Run a single plate or scale up to multiple plates with workflow options that support batching.
Integrated user interface
Touchscreen and command console are built into the instrument—no external dock station required with Smart Help features offered directly on the instrument.
Reliability
Engineered for consistent, long-term operation
SwiftArrayStudio is built as a robust research platform designed to support uptime and reproducible performance in high-throughput environments.
Precision plate handling
A novel autoteach capability for the plate gripper helps reduce positioning calibration errors.
Optimized fluid handling
On-platform placement of four 1 L ergonomic wash bottles with simple snap-in mounting helps reduce tubing and simplifies maintenance.
Integrated peristaltic pump
Supports controlled and reliable liquid movement throughout the system.
Long-life illumination system
An integrated multi-wavelength LED light source, designed to last, eliminates the need for periodic light source replacement.
Applications
Support
The SwiftArrayStudio Analyzer is supported by Thermo Fisher Scientific's comprehensive services and support to keep the system performing at its best. From installation, training, and digital support to remote troubleshooting and extended service plans, every stage of ownership is covered. These services help minimize downtime, ensure reliable performance, and provide long-term confidence in your investment.
Specifications
Attribute Specification
Dimensions (W x H x D) 54.52 x 25.03 x 26.92 in. (138.48 x 63.58 x 68.39 cm)
External waste bottle: 6 x 10 x 8 in. (15.24 x 25.4 x 20.32 cm)
Weight 375 lb (170.1 kg)
Temperature range 15°C–30°C (59°F–85°F)
Humidity range 20%–80% relative humidity, noncondensing
Pollution degree Pollution degree 2: Install the instrument in an environment that normally only has nonconductive pollutants, such as dust particles or wood chips. Typical environments are laboratories and sales and commercial areas.
Operating altitude Located between sea level and 6,500 ft. (2,000 m) above sea level
Electrical supply 100–240 VAC
Current 6.2–2.6 A
Main supply voltage fluctuations ±12% VAC
Illumination LED light source
Pixelation 4.1 megapixels
Scan time, 96-array plate 105 minutes
Scan time, 384-array plate 95 minutes
Instrument throughput utilizing automated array processing
Configuration Scan time—entire plate Scan time—per sample Max samples per week*
96-array plate 105 minutes 65.6 seconds 1,536
384-array plate 95 minutes 15 seconds 6,144
*Assumes a five-day work week
For Research Use Only. Not for use in diagnostic procedures.