Electron backscatter diffraction reveals the microstructure of metals and alloys
EBSD finds significant application in the study of metals and alloys, offering advantages over conventional techniques by providing a wide range of characterization possibilities in a single method. The microstructure of metals, alloys, and composites plays a crucial role in determining their desired properties, making EBSD an essential tool for their characterization and understanding the impact of different processing steps.
In manufacturing processes like joining, deformation, powder metallurgy, and additive manufacturing, EBSD is a powerful technique for characterizing and evaluating the influence of parameters and production stages on the final microstructure of components. It is also widely used to assess the effects of surface heat treatments and in situ mechanical tests.
Steel manufacturing
Specific industrial applications in which EBSD is regularly employed to enhance material quality and production processes
Aluminum sheets used for beverage cans or food containers require controlled texture to ensure smooth surfaces and sufficient strength. EBSD is utilized to assess and optimize the texture, ensuring the production of high-quality cans with reduced material usage.
In the automotive industry, the latest generation of high-strength steels is employed to reduce vehicle weight and improve fuel efficiency. EBSD helps differentiate between different phases (austenite, ferrite, martensite) and provides grain size and texture information crucial for optimizing the strength and performance of these steels.
Material quality and production processes
EBSD is widely used to understand failure mechanisms in metals. It can identify grain boundaries that are susceptible to corrosion and subsequent fracture. Grain boundary engineering (GBE), enabled by EBSD, can be employed to improve material properties by manipulating grain boundaries and enhancing corrosion resistance, creep resistance, and deformation behavior.
EBSD facilitates the study of dislocations and defects in metals, providing insights into deformation processes and their effect on mechanical properties. This information is particularly valuable for optimizing processing conditions, such as heat treatment and cold working, in materials like aluminum alloys, where corrosion resistance and lightweight properties are crucial.
Steel corrosion
Electron backscatter diffraction articles
How does EBSD work?
What is electron backscatter diffraction (EBSD)?
Combined EBSD and EDS Analysis
Thermo Scientific Apreo ChemiSEM System
The Apreo ChemiSEM System delivers all-around nanometer or sub-nanometer resolution at an analytical working distance of 10 mm and features fully integrated EDS and EBSD. Its field emission gun (FEG) source makes it possible to analyze a wide range of beam-sensitive materials, composites, magnetic samples, and nano-materials.