Search
Having difficulties with your experiment?
We are dedicated to your success. Get back on track. View our expert recommendations for commonly encountered problem scenarios.
View the relevant questions below:
Beginning your experiment?
Visit our
We typically recommend dislodging Sf9 cells mechanically rather than enzymatically (i.e., with trypsin/EDTA). To do so, wrap your flask with a towel, and bang the narrow end along the hood. This should cause the cells to dislodge from the flask.
This is normal for Sf9 cells. Typically, we recommend leaving the cells for 1 week after recovery, then checking cell viability. There should be approximately 1–2 million cells/mL, and can be split to a lower density.
These cells were oversplit. Typically, we recommend the lowest density for the Sf9 cells to be 0.5 million cells/mL.
Yellow particles could be cell organelles, aggregates, or debris. We see this when we first thaw frozen cells. To avoid this, you can let the shaker culture sit for 5 minutes, then transfer top 1/3 to a new flask, making sure to count cells first. You can also use heparin at up to 200 U/mL to decrease aggregation. Pluronic™ solution at a final concentration of 0.2% can also be used to decrease shearing, and increase shake speed to 100–120 rpm. Recently thawed cells seem to be breaking up and releasing small vesicles, as observed under high magnification. To reduce the amount of those small particles, cells need to be rapidly but completely defrosted for successful thawing to take place. Also:
Lastly, cells should be checked for contamination. To do so, plate a small portion of culture in a T-25 flask and incubate for 3 days, checking for cloudiness.
The cells are most likely getting too confluent. High Five™ cells grow extremely fast in serum. Split the cells as soon as there is no more room on the flask for the cells. We suggest doing a 1:15 split and observing cell growth each day. Add heparin at 10 μg/mL, although it can be used at 20 times higher concentration. Try adding some FBS (0.1%) for about 10–20 minutes then change the media.
These cells appear after cultures have been grown for several weeks. These do not seem to be detrimental to plating of high titer stocks or expression. However, if they are in high numbers, it may indicate that the cells are becoming old and that the culture should be re-started with a new stock of frozen cells.
For Research Use Only. Not for use in diagnostic procedures.