Automate your entire sample preparation workflow with the EXTREVA ASE Accelerated Solvent Extractor. The EXTREVA ASE system combines and automates sample extraction, in-cell cleanup, and evaporation. Faster extraction, with four samples extracted in parallel, increases laboratory throughput and efficiency, and the patented gas-assisted extraction mechanism uses less solvent and produces less waste. Designed with components that minimize PFAS background, this system provides greater accuracy and sensitivity for PFAS and other POPs testing, ensuring you achieve more precise results in your analyses.
With the EXTREVA ASE system, prepare your solid and semi-solid samples for GC, GC-MS and LC-MS analyses with ease. Simply place your sample cells into the extraction rack, set the desired parameters on the user interface and let the system take care of the rest. When extraction is complete, the EXTREVA ASE system automatically transfers your extracts to the evaporation carousel, eliminating the need for manual transfer of samples and additional sample preparation instruments. No other instrument or manual method encompasses the entire workflow without requiring user interaction or sample transfer at each step. The EXTREVA ASE system provides true walk-away operation for the entire process, making it the perfect partner for your GC, GC-MS or LC-MS analysis.
For labs that are extracting solid or semi-solid samples, the EXTREVA ASE system is the first fully automated sample-to-vial preparation platform. Common applications of the EXTREVA ASE system are to prepare samples for the analysis of:
  • Per- and polyfluoroalkyl substances (PFAS)
  • Organochlorine pesticides (OCPs)
  • Polycyclic aromatic hydrocarbons (PAHs)
  • Polychlorinated biphenyls (PCBs)
  • Dioxins
  • Furans
  • Fat content in food
  • Extractables & leachables (E&Ls) in biotherapeutics
The EXTREVA ASE system is thoughtfully designed to minimize PFAS background. This ensures more accurate and reliable results in PFAS analysis for environmental testing and compliance with local regulations. From environmental to food and beverage to pharma and biotech, the EXTREVA ASE system can be used in a variety of industries to prepare high-quality samples for GC- and LC-MS analysis.
The EXTREVA ASE system provides:
  • One seamless integrated operation—sample extraction, in-cell cleanup and evaporation
  • Increased throughput and efficiency—faster extraction with four samples in parallel
  • A green choice—patented gas assisted extraction mechanism saves up to 50% solvent usage
  • True walkaway technology—improves data quality and reduces errors at lower cost
  • Enhanced PFAS testing accuracy—built with components that reduce PFAS background
EXTREVA ASE system
* Required field
Style Sheet for Komodo Tabs
Style Sheet for Global Design System
CMD SchemaApp code